log in or register
MicroLog Temperature Data Logger EC600
EW-35710-91
$100.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
  • Compact, rugged housing guarantees operation in busy or rugged areas!
  • Automatically log temperature (°F or °C) readings for months, then download data to your computer Sample from once per 10 seconds to once per 2 hours—built-in clock and calendar track your data in real time
This logger stores up to 16,000 measurements, allowing you to track important temperature, and other process data for months. Logger is ideal for shipping, storage, laboratories, factories, refrigerators, freezers, incubators, desiccators, greenhouses, and more. The two-digit LCD shows current conditions—minimum and maximum readings are available at the touch of a button. Display flashes if set limits are passed.
Model 35710-91 records temperature and features a water-resistant and dustproof IP65-rated housing. Optional external sensors (sold separately) allow an additional parameter to be recorded. Connect current and voltage sensors with a transmitter to log other processes such as pressure, flow, pH, and level.
Software lets you set sampling rate, create graphs, and customize logging parameters. Software/cable package is required for use with loggers and are sold separately.35710-50 software/cable (sold separately) is required for use with 35710-00 Data Logger
Product Type Temperature Dataloggers
Range -22 to 122°F(-30 to 50°C)
Temp accuracy ±1.1°F(±0.6°C)
Temp resolution 1.0°F(0.5°C)
Display Two digit LCD display, 0.375 in H
Memory 16,000 data points
No. of channels 1 internal, 1 external
Battery life Approx. 2 years
Power 3.6 V lithium battery
Manufacturer number EC600
Model EC600
Accessories
Qty/Price
Availability
at $45.00 USD/EACH
Call 1-888-358-4705 for availability.
at $39.00 USD/EACH
In stock.
at $33.00 USD/EACH
In stock.
at $16.00 USD/EACH
In stock.
at $90.00 USD/EACH
In stock.
at $83.00 USD/EACH
Call 1-888-358-4705 for availability.
at $83.00 USD/EACH
Call 1-888-358-4705 for availability.
at $69.00 USD/EACH
Call 1-888-358-4705 for availability.
at $365.00 USD/EACH
Usually ships in 11 days.
Calibrations
Qty/Price
Availability
at $160.00 USD/EACH
Usually ships in 14 days.
Technical Resources
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title