Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Sorry, this item is now discontinued.browse similar itemsor Contact us for more information.
Micro Tri-Gloss Meter with RS-232 Output, 20/60/85 Degree
Item# EW-59721-17
$5,333.00 USD/EACH
Discontinued
Qty
  • Auto-calibration ensures high accuracy measurements
  • Evaluate the glossy surfaces of coatings, plastics, ferrous and nonferrous metals, and paper for quality control applications
  • Gloss meters 59721-02 through -17 feature auto-diagnosis— tells you when to calibrate and checks if standard is clean
  • Gloss meter models 59721-02 through -17 feature a scroll wheel and single button for easy operation.
Take these portable gloss meters with you anywhere—meters fit right in your pocket. Display single measurements or statistical functions including the number of readings, mean, standard deviation, min/max readings, range, difference, and pass/fail. Included easy-link software lets you easily transfer data from the meter to Microsoft® Excel® for quality control reports.
Checking Standards control the performance of the gloss meter and the surface condition of the calibration tile to guarantee accurate and repeatable measurements. Order separately.
What's included:
calibration holder and tile, software, RS-232 interface cable, one AA battery, and carrying case.
Product Type Gloss Meters
Output RS-232
Dimensions 6" W x 3" H x 2" D
(15.2 x 7.6 x 5.1 cm)
Qty/ea 1
Power 1 AA battery
Model AG-4446
Accessories
Qty/Price
Availability
at $9.25 USD/PACKAGE of 4
In stock.
at $2,048.00 USD/EACH
Usually ships in 10 days.
Technical Resources
Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
Questions and Answers
Do you have questions about this product?
Get answers from real customers and in-house experts with AnswerBox.
1 Question | 1 Answer
Displaying question 1
  • Q:
    Hi,
    I am interested in measuring the reflectivity of surfaces. What is the range of gloss values? Can this tool be used to measure reflectrivity or emissivity of solid surfaces (flat and curved)?
    Thanks,
    Murugesu Yoganathan
    Asked on 8/14/2013 by Yoga from NY
    1 answer
    • STAFF EXPERT
      A:
      This product measures gloss in the range of 0-2000 gloss units (GU). It can measure both matte and high-gloss substrates, but flat surfaces will provide the best readings. Emissivity is a different type of measurement, but is also something that we can provide.
      Answered on 8/15/2013 by Jack from Technical Support
Displaying question 1
Do you have a question about this product? 
Product Reviews