United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 2
of
2
EW-06274-11
MICROCENTRIFUGE TUBE ICE RACK (HOLDS 120 X 1.5 ML TUBES)
Click to enlarge
More Product Details
Specifications
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-06274-11
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Laboratory Supplies & Consumables
>
Ice Buckets and Ice Pans
>
Microcentrifuge Tube Ice Tray, polystyrene
Share
|
Microcentrifuge Tube Ice Rack (holds 120 X 1.5 mL Tubes)
EW-06274-11
$87.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
For temperature stabilized benchtop ice storage
These two piece racks are molded of high-impact polystyrene with a lower section tray that holds water or ice flakes. Model 06274-09 is stackable and holds 105 400 µl microcentrifuge tubes, or other tubes with a diameter up to 8 mm. Model 06274-11 is not stackable and holds 120 1.5 ml microcentrifuge tubes, or other tubes with a diameter up to 12 mm.
Specifications
Product Type
Centrifuge/Microcentrifuge Racks
Tube size
1.5 ml microcentrifuge tubes or other tubes with a diameter under 12 mm
Tubes held
120
Material
Polypropylene
Manufacturer number
H18907-0000
Model
H18907-0000
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title