Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
 

Microplate reader, 340 to 750 nm, 6, 12, 24, 48, 96 well

EW-13055-54
$6,055.00 USD/EACH

Call 1-888-358-4705 for availability.
Qty

  • Optional 72/96 Terasaki and 3840well capacity, UV 340nm capability
  • Stores up to 75 custom assay definitions and 8 test results
  • Extensive onboard data analysis and optional software which offers more flexible data manipulation
The Elx800 microplate readers feature on board data reduction and reporting. Reads 6-, 12-, 24--, 48-, and 96-well plate microplates. The model 13055-52 can read 384-well and 60-, 72- and 96-well Terasaki plates. Wavelength range from 400 to 750nm. Model 13055-54 optics to extend wavelength range down to 340nm.

Microplate readers feature a 2x24-character LCD and convenient soft-keys. Extensive curve fitting options include linear, cubic, quadratic, 4-p, log-logit, cubicspline, and point-to-point. Memory stores 25 standard curves and up to 8 microplate test results. Features auto or manual plate layout for blanks, controls, standards, and samples. Also offers control and assay validation, transformation and formula applications, and cutoff and call criteria.

Onboard software is easy to program to run sophisticated assays and analyzed without the need of computers. Additional software 13055-56 offers easy user interface with flexible data manipulation and curve fitting for basic endpoint, kinetic, multi-wavelength and spectral scan requirements. Software package includes software on CD, and three user licenses. What’s included: 340 (13055-54 only), 405, 450, 490, 630nm filters, dust cover, parallel printer cable (25-pin male and 36-pin male), serial cable (null modem, 9-pin female to 25-pin female), operator’s manual. Additional filters are available range from 304 to 750nm. Call our application specialist or visit our web site for more information.

Accuracy (UV range (340 nm)) <1% at 2.0 OD
Accuracy (Visible range (400-750 nm)) <1% at 2.0 OD
Absorbance range (UV range (340 nm)) No
Absorbance range (Visible range (400-750 nm)) Yes
Linearity 1% from 0 to 2.0 OD
Wavelength range 340 to 750 nm
Minimum plate read time 96 wells in 30 second
Light source tungsten halogen bulb
Power 100/240 V, 50/60Hz
Dimensions 15x7x16.5 (38.4x17.8x41.9 cm)
Manufacturer number ELX800UV





Technical Resources
 
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.

See Details
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title