United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
16 >
EW-07105-00
Mini Submersible Magnetic Drive Pump, 3.0 GPM, 115 VAC
Click to enlarge
More Product Details
Specifications
Technical Resources
Flow Charts
Instruction manual
Virtual Catalog (PDFs)
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-07105-00
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Pumps
>
Submersible Pumps and Sump Pumps
>
Mini Submersible Magnetic Drive Pumps
Share
|
Mini Submersible Magnetic Drive Pump, 3.0 GPM, 115 VAC
EW-07105-00
$202.00 USD/EACH
Usually ships in 5 days.
Qty
:
Add to Cart
Save to shopping list
Operate fully submerged or in open air
Good for low-viscosity liquids including solvents, dyes, inks, and mild corrosives
Liquid-cooled models keep heat-sensitive fluids from warming up
Thick (1/8") epoxy coating dissipates heat quickly and provides good chemical resistance
Also includes a plug.
What's Included:
A three-wire cord.
Specifications
Product Type
Submersible Centrifugal Pumps
Max flow rate (GPM)
3
Max flow rate (LPM)
11.4
Max head (ft)
4.6
Max head (m)
1.4
Max system pressure (psi)
50
Max system pressure (bar)
3.4
Motor (rpm)
3200
Motor (VAC)
115
Motor (Hz)
60
Motor (Amps)
0.45
Motor (hp)
1/200
Motor (type)
TENV
Wetted materials
PP head, glass-filled nylon impeller, Buna-N O-ring, 316 SS spindle
Max fluid temp (º F)
190
Max fluid temp (º C)
87
Duty cycle
Continuous
Connections (inlet)
1/2" OD
Connections (outlet)
1/2" OD
Submersible
Yes
Dimensions
4-1/2"L x 3-3/4"W x 3-1/4"H
(11.4 x 9.5 x 8.3 cm)
Technical Resources
CP_Flow_Curve_07105-00.pdf
Instruction Manual (154 KB)
Virtual Catalog (PDFs)
13-14 Catalog
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title