Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
 

Miniature Laser Alignment Kit with Dot Image; No Accessory Lens, 500 ft Max Range

EW-41935-00
$219.00 USD/EACH

Usually ships in 6 days.
Qty

  • Projects laser dot image up to 1000 feet
  • Small size is ideal for tight areas—mount to any 1" x 1" flat surface.
Laser features adjustable focus and straight-line alignment to 0.0156" over entire beam length. Order additional optics for more projection images.
What's included: a miniature laser diode module with adjustable focus, an X-Y adjustable mounting tube, L-shaped mounting bracket, accessory optics, and AC adapter.
Range 500 ft
Max output <= 5 mW
Wavelength 650 nm
Class IIIa
Dot size 1/32" at 3", 1/16" at 5 ft, 1/8" at 25 ft, 1" at 150 ft
Dimensions 1 3/4"L x 7/8"W x 1"H (without accessory lens)
Power 5 VDC or 110 VAC (AC adapter included)





Technical Resources
 
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title