Miniature gear pump, 0.67 GPM (2520 mL/min), 12 VDC lab model
YO-07012-20
$59.00 USD/EACH
Are you a reseller or buying in bulk? Call us today.
  • Complete mini gear pump systems for low-flow applications
  • Maximum flow rates up to 0.67 GPM
  • Choose from nonenclosed OEM, lab, and fully enclosed lab versions
  • 115 VAC, 230 VAC, and 12 VDC models
What's Included:
models 07012-00, -02 have 12" (30.4-cm) leads with no plug; models 07012-30, -32 have an aluminum enclosure and a 7-ft (2.1-m) cord with grounded plug and in-line switch; model 07012-20 has an 8-inch (20.3-m) cord with no plug.
Wetted parts DELRIN® plastic, 304 stainless steel, Buna N, and PTFE®Acetal
Power (VDC) 12
Max pressure 21 psi
Max temperature 200°F (93° C)
Max flow rate (Liters/min) 2.52
Max flow rate (GPM) 0.67
Port size 3/16" hose barb
Particulates No
Run dry No
Relief valve No
Suction lift (dry) 4"
Suction lift (wet) 10 ft
Duty cycle Continuous
Viscosity 200 cp max
Motor (Amps) 2.0
Power cord 8"cord, no plug
Dimensions 4"Lx1-3/4"Wx2-5/8"H
Technical Resources
Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Do you have questions about this product?
get answers from real customers and in-house experts with AnswerBox.
48 Questions | 49 Answers
Displaying questions 1-10Previous | Next »
  • Q:
    What is the expected life at 50% duty cycle in an 8 hr per day closed loop water cooling system. Max temp around 60C
    Asked on 3/19/2014 by Dave from Warren OH
    1 answer
    • STAFF EXPERT
      A:
      The life of the motor is roughly 1000 hours
      Answered on 3/20/2014 by Anthony from Technical Support
  • Q:
    whats the minimum flow rate?
    Asked on 2/5/2014 by Erttu
    1 answer
    • STAFF EXPERT
      A:
      The flow rate on this pump can be controlled by varying or restricting the voltage supplied to the pump. There is not a specified minimum flow rate.
      Answered on 2/6/2014 by Dan from Technical Support
  • Q:
    Is it self-priming?
    Asked on 1/22/2014 by Shel from la jolla
    1 answer
    • STAFF EXPERT
      A:
      This is not a self-priming pump.
      Answered on 1/22/2014 by Anthony from Technical Support
  • Q:
    is the flow on this pump reversible and will it work in a miniature hydraulic system.
    Asked on 10/27/2013 by ronald from temecula ca.
    1 answer
    • STAFF EXPERT
      A:
      The pump flow is reversible, and it could work in a miniature hydraulic system, so long as the hydraulic fluid is compatible with the pump.
      Answered on 10/28/2013 by Jack from Technical Support
  • Q:
    The product calls for a 12V DC power supply however it doesn't specify the Amperage. What specifications should an accompanying power supply include?
    Asked on 10/10/2013 by Brent
    1 answer
    • STAFF EXPERT
      A:
      The maximum is 2.2 amps.
      Answered on 10/11/2013 by Dan from Technical Support
  • Q:
    Is this pump CE marked?
    Asked on 7/17/2013 by Peter from Nottingham, UK
    1 answer
    • STAFF EXPERT
      A:
      This pump is not CE marked.
      Answered on 7/17/2013 by Polo from Technical Support
  • Q:
    Hi
    Will this work for drain out the waste for chemical solvent like Paraffin, Alcohole
    Thanks
    Kishor
    Asked on 4/3/2013 by Kishor from Hyderabad, India
    1 answer
    • STAFF EXPERT
      A:
      The solvents listed would not be compatible with the Buna-N seals in the pump. Use with these chemicals may also require the use of an intrinsically safe or explosion-proof pump. Please contact our technical department for further discussion regarding this procedure.
      Answered on 4/3/2013 by James from Technical Support
  • Q:
    Does this pump have a smooth flow?
    Asked on 2/25/2013 by sun from cincinnati, OH
    1 answer
    • STAFF EXPERT
      A:
      Yes, gear pumps have a pulseless, smooth, flow.
      Answered on 2/26/2013 by Jen from Technical Support
  • Q:
    I am looking to use this pump in a student unmanned submersible project to move water for ballast. If the power shuts off will water flow through the pump? Do I need a valve to prevent water flowing through the pump when it's not in operation?
    Thanks,
    Nate
    Asked on 10/17/2012 by Nate from Baltimore, MD
    1 answer
    • STAFF EXPERT
      A:
      Unit is not water tight, not recommended to be submersed in any fluid.
      A check valve on the outlet may be needed to prevent back-flow.
      Answered on 10/19/2012 by Jen from Technical Support
  • Q:
    can this pump handle both methanol/nitromethane mix as well as gasoline? I know it is not rated for flammable liquids, however this is used in an outdoor, open environment.
    Asked on 9/17/2012 by Jeff from Virginia
    1 answer
    • STAFF EXPERT
      A:
      The Buna N (nitrile) from the wetted materials list is not compatible with the nitromethane component, so we do not recommend using this pump with that mixture. The electronics are not sealed, so there is a spark hazard. Even in an outdoor environment we would not suggest using this pump with a source of flammable vapors nearby.
      Answered on 9/19/2012 by Christina from Technical Support
Displaying questions 1-10Previous | Next »
Do you have a question about this product? 
Product Reviews
REVIEW SNAPSHOT®
by PowerReviews
Miniature gear pump, 0.67 GPM (2520 mL/min), 12 VDC lab model
4.5
(based on 4 reviews)
100%
of respondents would recommend this to a friend.
Pros
  • Works properly (4)
  • Durable (3)
  • Easy to use (3)
Cons
    Best Uses
      Reviewed by 4 customers
      Sort by
      Displaying reviews 1-4
      Back to top
      4.0
      Pump runs well.
      By The Kid
      from Northern Kentucky
      About Me Student
      Verified Buyer
      Pros
      • Accurate
      • Durable
      • Easy To Use
      • Works Properly
      Cons
        Best Uses
          Comments about Miniature gear pump, 0.67 GPM (2520 mL/min), 12 VDC lab model:
          used in greenhouse on solar heating system.
          4.0
          Works Great, but noisy
          By Ed
          from North Carolina
          About Me Academic/Professional Researcher
          Verified Reviewer
          Pros
          • Durable
          • Easy To Use
          • Powerful
          • Reversable Polarity
          • Variable output
          • Works Properly
          Cons
          • Bit Noisy
          Best Uses
            Comments about Miniature gear pump, 0.67 GPM (2520 mL/min), 12 VDC lab model:
            I am using it to circulate hot water in home.
            Connected with PEX tubing that fits the barb fitting and standard 3/8 compression fitting.
            It has stalled, jammed, or failed to start pumping at 6volts but seems very reliable to start at 8 volts and up. The motor got very hot when it stalled, but fortunately did not burn out. It seems to be built very robust.
            Impressed with the pressure output and inlet draw.
            The only negative is the noise level.
            Have not measured the decibels, but an irritating rather loud hum at the speed I chose to run it.
            Good value, well performing and satisfied purchase.
            (1 of 1 customers found this review helpful)
            5.0
            works great!
            By John
            from Michigan
            Verified Buyer
            Pros
            • Works Properly
            Cons
              Best Uses
              • In The Field
              Comments about Miniature gear pump, 0.67 GPM (2520 mL/min), 12 VDC lab model:
              Works great!
              (1 of 2 customers found this review helpful)
              5.0
              Good pump for low flow applications
              By Matt
              from Lakewood, CO
              About Me Academic/Professional Researcher
              Verified Buyer
              Pros
              • Durable
              • Easy To Use
              • Works Properly
              Cons
              • Short electrical leads
              Best Uses
              • In The Field
              Comments about Miniature gear pump, 0.67 GPM (2520 mL/min), 12 VDC lab model:
              I use the product for an anaerobic bioreactor. The pump works great, and is incredibly durable. The pump can be connected to a variable power supply and control the flow rate.
              Displaying reviews 1-4
              Back to top
              Customers Who Bought This Item Also Bought
              View products by: Title | Brand | Category | Price | Popularity | Best Sellers
              View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
              View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
              View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title