United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
2 >
EW-75420-00
Moderate Flow Positive Displacement Bypass Diaphragm Pump, 1.4 GPM, 115 VAC
Click to enlarge
More Product Details
Specifications
Technical Resources
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-75420-00
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Pumps
>
Positive Displacement Pumps
>
Diaphragm Pumps
>
Bypass Diaphragm Pumps
Share
|
Moderate Flow Positive Displacement Bypass Diaphragm Pump, 1.4 GPM, 115 VAC
EW-75420-00
$138.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Ideal for spray nozzles and transfer applications where smooth, consistent flow is required
Pumps accommodate any size spray orifice. Maintain a near-constant pressure across pumping range for any size nozzle used. A flexible diaphragm design allows pump to run dry without damage.
What's included:
115 VAC model includes a 6-ft power cord: 12 VDC model has 6" leads.
Specifications
Product Type
Diaphragm Pumps
Wetted materials
Polypropylene body, Santoprene® diaphragm, Viton® valves
Port size
3/8" NPT(F)
Max flow rate (LPM)
5.3
Max pressure
45 psi
Max temperature (º C)
60
Max temperature (º F)
140
Run dry
Yes
Self-priming
8 ft
Particulates
No
Power (Amps)
0.51
Power (Hz)
60
Power (VAC)
115
Duty cycle
Continuous
Dimensions
6-1/2"L x 3-3/8"W x 4"H
Technical Resources
Related technical articles (Roll over for summary)
Positive Displacement Pump Types
How to Read a Pump Curve
Guide to Liquid Pumps
Show All...
Liquid Pump Installation Information
Liquid Pump Selection Flowchart
Liquid PumpsViscosity Handling Characteristics
Motor Enclosure Terminology
InnoCal Pump Reconditioning Program
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title