United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 4
of
9 >
EW-67035-10
Modular stainless steel drying rack, 50 white pegs
Click to enlarge
More Product Details
Specifications
Accessories
Build a complete system
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-67035-10
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Laboratory Supplies & Consumables
>
Labware
>
Laboratory Organizers
>
Labware, Racks & Supports
>
Modular Drying Racks, 304 Stainless Steel
Share
|
Modular stainless steel drying rack, 50 white pegs
EW-67035-10
$581.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Removable "snap-in" pegs and add-on accessories let you turn these drying racks into customized work stations in just seconds!
Corrosion-resistant 304 stainless steel (SS) racks and polypropylene pegs are easy to clean.
Autoclavable.
Integral catch drain directs all drainage into the drip troughprevents accidental drainage to the back of the rack.
Racks come with 6"L pegs, SS drain tube, and wall mounting bracket. Order add-on accessories, plugs, and additional pegs separately.
Assorted peg colors include yellow, orange, pink, red, green, light blue, dark blue, lilac, and purple.
Specifications
Product Type
Drying Racks/Stands
Material
Stainless Steel
Dimensions
30"W x 30"H x 4"D
Width (in.)
24
Width (mm)
610
Build a complete system
1
.
Modular Drying Racks, 304 Stainless Steel
Details
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Physical Properties of Plastics
Physical Properties of Glass
Physical Properties of Stainless Steel
Show All...
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title