United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 4
of
14 >
Representative photo only
Multiwash III Plate Washer with 8 Port Coaxial Manifold
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-13054-14
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Life Science
>
Immunochemistry
>
Microplate Products
>
Microplates
>
Microplate Washers
>
Multiwash III Plate Washers
Share
|
Multiwash III Plate Washer with 8-Port Coaxial Manifold
EW-13054-14
$4,624.50 USD/EACH
Usually ships in 7 days.
Qty
:
Add to Cart
Save to shopping list
Rinse-on-demand feature for complete system cleaning at the touch of a button
Quick-connect fittings allow unit to be fully operational in minutes
Can handle full range of assay types from the most gentle wash to the most aggressive ELISA
Ideal for labs performing a variety of routines including -- agglutination tests; cytotoxicity tests; and ELISA, EIA, FIA, and LIA tests. Washer features easy-to-program wash routines, bottom washing, sweeping crosswise aspiration, dispense only and aspirate only functions, and overflow washing for continual dispensing while aspirating off the top for a thorough wash. By programming and storing routines, you can switch between seven different modes at the touch of a button.
The washer performs a 300-µL three-cycle wash on a full 96-well plate in less than two minutes. Washer is can run up to ten wash cycles. You control the dispense pressure and soak time. The washer also features a floating manifold that adjusts to match each plate and allows the washer to perform special test procedures. Extra aspiration adjustment for both horizontal and vertical needle positions can be stored in individual programs.
Independent adjustments of horizontal and vertical allow any shape well to be accommodated. Washer has a wash and rinse reservoir volume of two liters each. Wetted materials include glass, nylon, PTFE
®
, stainless steel, polypropylene, Tygon
®
, Delrin
®
, polyethylene, and Santoprene
®
. Measures 11'W x 81/2'H x 145'D. All units are also certified with CAN/CAS std c 22.2 No. 1010.1.
What's Included:
manifold (except 13054-34); slip connectors; reservoir kit; fuse; rinse strips; dust cover; operation manual; quick-start guide, and two power cords (U.S. standard and European). Order additional manifolds separately.
Specifications
Product Type
Plate Washers
Full plate processing time
180 seconds
Soak time
0 to 999 seconds
Wash cycles
10
Accuracy
+/- 3% at 300 uL
Brand
Multiwash
Manufacturer number
820-04(MULTIWASH3)
Accessories
Qty
/
Price
Availability
8-Port Coaxial Manifold for Multiwash III Plate Washers
EW-13054-26
at
$331.00 USD/EACH
Usually ships in 7 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Technical Expertise: How Cole-Parmer Application Specialists Respond to Customers' Needs
Isolating Genomic DNA from Whole Blood
Effects of Mounting a Breathable Membrane on the 384-Well Plate
Show All...
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title