Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Myron L Soil pH and Conductivity Meter Kit

Item# EW-99040-00

$749.00 USD/EACH

Are you a reseller or buying in bulk? Call us today.


Usually ships in 3 days.
Qty

  • Save time and money--test your soil on location!
  • Cup-style sensor features self-maintaining conductivity electrode
  • Monitor soil pH, conductivity, alkalinity for better plant health
Plant health and productivity is greatly affected by the pH, salinity, and alkalinity levels of your soil. These vital factors are often overlooked by many growers resulting in poor plant health. This new soil pH and conductivity meter allows you to easily test and monitor these factors so that plants are healthier and more productive.
Use this hand-held meter in the field. Simply take a small soil sample, mix with water after the alloted time, pour the sample water into your meter, and press the black button for conductivity readings, or the red button for pH readings. Alkalinity testing is just as simple! Read alkalinity in ppm by comparing the solution level to the scale on the side of the test vial.
Product Type Soil test kit
Display analog dial
Conductivity range 0-5 millimhos
pH range 2-12 pH units
pH accuracy ±0.2 pH units
Conductivity accuracy ±2% of full scale
Repeatability ±1%
Temp compensation automatic to 25°C
Dimensions 3-1/2"W x 4.0"H x 4-1/2"D
Battery life approx. 2000 tests
Ambient conditions 50 to 160°F (10 to 71°C)
Power Two 9 V batteries (included)
Qty/ea 1
Brand Mryon L

Accessories
Qty/Price
Availability

at $13.00 USD/PINT

In stock.

at $13.00 USD/PINT

In stock.

at $13.00 USD/PINT

In stock.

at $167.00 USD/EACH

Usually ships in 3 days.

at $39.00 USD/QUART

In stock.
Calibrations
Qty/Price
Availability

at $126.00 USD/EACH
Usually ships in 6 days.




Technical Resources
 
Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
Questions and Answers
Product Reviews