InnoCal NIST-Traceable Calibration; Hydrometer, 1-point
Representative image only.
InnoCal Item # EW-17107-00
NIST-traceable calibrations for hydrometers
  • Provides traceability to NIST
  • Lists the environmental conditions present at time of calibration
  • Certificate lists actual values, uncertainties, and tolerances
  • Regular calibration helps you meet ISO, EPA, GLPs/GMPs and other quality standards
  • A2LA accredited
PRODUCT OPTIONS
$176.00USD / Each
Often Bought with…
SPECS & DESCRIPTION
Specifications & Description
  • Calibration TypeLiquid Testing
  • System Calibration (Instrument and Probe)Yes
  • Number Of Calibration Test Points1
  • Calibration Test PointsMid-Point of the range
  • Calibration Test Range0.65 to 2.0 specific gravity
  • DescriptionNIST-Traceable Calibration; Hydrometer with test data, one-point
Key Features
  • Provides traceability to NIST
  • Lists the environmental conditions present at time of calibration
  • Certificate lists actual values, uncertainties, and tolerances
  • Regular calibration helps you meet ISO, EPA, GLPs/GMPs and other quality standards
  • A2LA accredited
More About this Item
All calibration certificates contain as-found and as-left data, estimated measurement uncertainties (emu’s) and test uncertainty ratios (TUR’s) at no additional cost!
NIST-traceable calibration allows you to confirm that the specific asset you purchase does in fact meet the manufacturer’s published specification. Receive your product calibrated and ready to use rather than sending it to a third-party or internal calibration lab and incurring additional delays. EPA, USDA, ISO and GLP/GMP standards require regular NIST-Traceable calibration of all test equipment being used in critical applications.