log in or register
Needle Valve, 304 Stainless Steel, 5000 Max psi
EW-68831-10
$80.00 USD/EACH
Usually ships in 6 days.
Qty
    Use these needle valves to closely regulate a fine flow. You can also use these valves as a throttling device on lines in which rapid or excessive pulsing pressures effect gauge performance. Connections are 1/4'' NPT(F).
    Product Type Pressure gauge accessories
    Wetted materials 304 SS
    Range 0 to 5000 psi
    Process connection 1/4" NPT(F)
    Technical Resources
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title