United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
Representative photo only
Nilfisk Backpack HEPA Vacuum 120 VAC
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-86564-20
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Clean Room
>
Vacuums
>
Nilfisk Backpack HEPA Vacuum
Share
|
Nilfisk Backpack HEPA Vacuum, 120 VAC
EW-86564-20
$707.00 USD/EACH
Usually ships in 8 days.
Qty
:
Add to Cart
Save to shopping list
Lightweight, ergonomic design to reduce fatigue
Take this compact vacuum wherever you need it! Backpack style allows you to clean tighter areas where conventional vacuums won't fit. Low noise level of 62 dB at 1 meter increases operator comfort. HEPA filter traps 99.99% of particles down to and including 0.3 microns. Exhaust air is directed upwards so that it doesn't interfere with the operator.
What's Included:
telescoping wand, 6-ft (1.8-m) vacuum hose, round brush nozzle, combination floor/rug nozzle, crevice nozzle, carrying harness, filters (main, exhaust, HEPA), five dust bags, and 50-ft (15-m) extension cord.
Specifications
Product Type
Cleanroom Vacuums
Capacity
2.6 gal. (9.8 L)
Suction lift
96 in (244 cm)
Air flow rate
139 cfm
Power (VAC)
120
Dimensions
23"H x 9-3/8"OD (58.4x 23.9)
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title