dummy
EW-06645-80
Filter membranes are ideal for use with the gel dryer or semidry blotter. Use pure nitrocellulose membranes for nucleic acid and protein transfers, plaque lifts, and filter hybridization. Binding capacity is 80-100 µg/cm2. Use positively charged nylon membranes for RNA and low-molecular weight DNA transfers without high salt buffers, or whenever a low-protein bonding surface is desirable. Use for multiple DNA or RNA reprobings (not recommended for protein sampling). Nylon membranes are tear-resistantwill not crack or curl.
This item is considered a hazardous material.Please note: there may be additional shipping restrictions like freight collect or no priority air service, extra freight charges may apply. This item(s) may be restricted from shipping to a residential address, sold to private individuals or exported from the United States. For additional information or questions please call us at 1-800-323-4340 (US) or 847-549-7600 (International).
| Product Type |
Nitrocellulose Membranes |
| Filter size |
0.45 um |
| Membrane support |
Nitrocellulose |
| Applications |
DNA/RNA transfers/reprobing |
| Pore size (micron) |
0.45 |
| Dimensions |
300 cm x 33 cm |
at $260.00 USD/BOX of 100
Call 1-888-358-4705 for availability.
at $205.00 USD/BOX of 100
Call 1-888-358-4705 for availability.
Technical Resources
Related technical articles (Roll over for summary)
Show All...
Show Less...
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334