Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

North Full-Face Respirator, Thermoplastic elastomer

EW-81606-22
$137.00 USD/EACH

  • Wraparound lens provides over 200° field of vision
  • Superior comfort and fit
  • Contoured flange ensures proper seal
Soft, pliable silicone and economical thermoplastic elastomer models, both feature hard-coated, scratch-and impact-resistant wraparound polycarbonate lens providing over 200° field of vision. Lens protects eyes and face against irritating gases, vapors and flying particles. The oral/nasal cup with dual sealing flange reduces fogging. Silicone face piece features a speaking diaphragm for clear communication.
Order filter cartridges according to demands of work environment (cartridges sold separately).
Compliance: ANSI Z87.1-1989
Approval: NIOSH
Product Type Full-face respirators
Compliance ANSI Z87.1-1989
Color Black
NIOSH Approval NIOSH
Brand North
Manufacturer number 54001

Accessories
Qty/Price
Availability

at $16.50 USD/PACKAGE of 2

In stock.

at $18.00 USD/PACKAGE of 2

In stock.

at $30.00 USD/PACKAGE of 2

In stock.

at $30.00 USD/PACKAGE of 2

In stock.




Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title