Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

North Half-Mask Respirator, Medium

EW-81606-12
$33.00 USD/EACH

  • Wraparound lens provides over 200° field of vision
  • Superior comfort and fit
  • Contoured flange ensures proper seal
These half-mask respirators are made of soft, pliable, non-allergenic silicone offering excellent comfort and fit. Low-profile positioning on the face gives a wide field of vision and room for protective eyewear. Contoured, extended side sealing flanges and design of nose area provide a good seal for easy communication or other facial motions. Removable cradle-suspension straps offer a balanced, comfortable and secure fit.
Approval: NIOSH
Product Type Half-mask respirators
Color Black
NIOSH Approval NIOSH
Size Medium
Brand North
Manufacturer number 770030M-12

Accessories
Qty/Price
Availability

at $16.50 USD/PACKAGE of 2

In stock.

at $18.00 USD/PACKAGE of 2

In stock.

at $30.00 USD/PACKAGE of 2

In stock.

at $30.00 USD/PACKAGE of 2

In stock.




Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    Customers Who Bought This Item Also Bought
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title