Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Nylon Row-Detail Brush, 7" Handle, 0.25"L x 0.5"W x 0.5"D Brush; 12/Pk

Item# EW-84551-35

$45.25 USD/PACKAGE

Are you a reseller or buying in bulk? Call us today.


    Use detail brushes for cleaning and dusting your delicate instruments. Also ideal for gently applying chemicals or working solutions into to intricate areas.
    Description 7 in Nylon Row Detail Brush
    Product Type Detail Brush
    Material Nylon Brush, Plastic Handle
    Qty/pk 12
    Color White
    Dimensions 2 x 8 Rows





    Technical Resources
    Virtual Catalog (PDFs)
  • Catalog - English
  •  
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
    Questions and Answers
    Product Reviews
    Customers Who Bought This Item Also Bought