Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Oakton 10.00 pH buffer, 4 L

EW-05942-64
$42.00 USD/EACH

quantity discounts Quantity Discounts
Order Discount Price
[X]
6 15% $35.70/EACH
    Oakton® buffers for meters with 0.01 resolution are standardized against NIST-traceable pH references within 0.01 pH at 77°F (25°C). Store buffers away from direct sunlight. Sold in individual bottles; 500-mL bottles are also available in cases of 12, 4-L bottles in cases of four.
    Product Type pH Buffers
    Description pH 10.00
    Volume (mL) 4000
    Quantity 4 L
    Brand Oakton
    Manufacturer number 05942-64





    Technical Resources
    Certificate of Analysis
    Enter lot number

    or click to view a Sample C of A
     
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    Customers Who Bought This Item Also Bought
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title