log in or register
Oakton CON 6+ handheld conductivity meter kit
EW-35604-05
$482.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
  • Autoranging conductivity with temperature measurement
  • CON 6+ meters have five auto-ranging measurement modes—up to 200.0 mS (200.0 ppt)
  • Temperature readout in °C with the push of a button
  • Rubber boot with built-in stand protects meter and adds functionality
These microprocessor-based meters feature easy, push-button operation that offers high accuracy at an extremely affordable price. A quick and easy single-point calibration covers all five ranges with the CON 6+ meters; for greater precision, single points can be calibrated within each of the five measurement ranges.
Additional features include a convenient Hold function and auto-off function that shuts meter off after 20 minutes of nonuse. Programming functions allow customization of meter for temperature compensation, temperature coefficient, normalization, and cell constant for better accuracy in varying applications.
What's Included:
protective rubber boot with stand, conductivity/temperature probe (K = 1.0), and four AAA batteries.
Kits include: protective rubber boot with stand, conductivity/temperature probe (K = 1.0),
calibration solutions, sample bottles, rinse bottle, hard plastic carrying case, and four AAA batteries.
Product Type Conductivity Meter
Meter style Portable
Dimensions (" W) 5-1/2
Dimensions (" H) 2-3/4
Dimensions (" D) 1-5/16
Dimensions (" L) 5-1/8 (probe)
Dimensions (" Dia) 5/8 (probe)
Range (Conductivity) 0 µS to 200 mS
Range (Temperature °C) 10 to 110
Range (Temperature °F) 14 to 230
Resolution (Temperature) 0.1
Resolution (Conductivity) 0.01, 0.1 µS; 0.001, 0.01, 0.1 mS
Temperature compensation Automatic or manual
Display LCD
Calibration Up to 5 points
CE Compliance Yes
Accessories
Qty/Price
Availability
at $21.25 USD/PACKAGE of 12
In stock.
at $159.00 USD/EACH
In stock.
at $155.00 USD/EACH
In stock.
at $159.00 USD/EACH
Call 1-888-358-4705 for availability.
Calibrations
Qty/Price
Availability
at $115.00 USD/EACH
Usually ships in 9 days.
Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  • More info
  • Instruction Manual for 35604 series
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title