log in or register
Oakton DO 2700 Benchtop Dissolved Oxygen Meter
EW-35416-00
$1,845.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
  • Designed for optimal performance and versatility
  • Advanced meter for research grade use with BOD capability
Oakton DO 2700 meter measures dissolved oxygen in mg/L, ppm, or % saturation with simultaneous temperature display in °F or °C. Large LCD features annunciators to aid in meter use and troubleshooting. Large display size makes viewing measurements easier. Meter is encased in a compact housing which takes up a minimum of bench space. Water-resistant membrane keypad and convenient pullout reference guide provide useful features for the lab.
Meter meets GLP requirements—all calibration and measurement data is stored with a time/date stamp. Bundled self-stirring probe and bi-directional RS-232 make this meter ideal for BOD and other advanced dissolved oxygen applications. Nonvolatile memory stores up to 500 data sets to simplify documenting data. Oversized graphic LCD with backlight allows readings under all conditions.
What's Included:
self-stirring dissolved oxygen/BOD probe, 6 membrane caps and electrolyte, and 100/240 VAC power adapter.
For complete specifications and operating instructions please view the instruction manual
Product Type Dissolved Oxygen Meter
Meter style Benchtop
Dimensions (" W) 6-7/8
Dimensions (" H) 2-3/4
Dimensions (" L) 6-1/8
Range (Salinity) 0 to 50 ppt
Range (Dissolved oxygen) 0 to 19.99mg/L
Range (Temperature °C) 10 to 110
Range (Temperature °F) 14 to 230
Resolution (Temperature) 0.1
Accuracy (Temperature) ±0.5°C
Temperature compensation Automatic or manual
Display LCD
Interface RS-232
Calibration Up to 6 points
Power 100 to 240 VAC, 50/60 Hz
Datalogging (points) 500
CE Compliance Yes
Brand Oakton
Accessories
Qty/Price
Availability
at $39.00 USD/EACH
In stock.
at $38.50 USD/EACH
In stock.
at $695.00 USD/EACH
In stock.
at $675.00 USD/EACH
Call 1-888-358-4705 for availability.
at $181.00 USD/EACH
Call 1-888-358-4705 for availability.
Calibrations
Qty/Price
Availability
at $115.00 USD/EACH
Usually ships in 16 days.
Technical Resources
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title