United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
3 >
EW-35625-15
Oakton
®
Food TempTestr
®
Infrared Thermometer
Click to enlarge
More Product Details
Specifications
Accessories
Calibrations
Technical Resources
Virtual Catalog (PDFs)
Brochure
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-35625-15
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17004-00
NIST-Traceable Calibration; IR Thermometer
$130.00 USD/EACH/EACH
Usually ships in 4 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Temperature, Humidity, Moisture and Weather Products
>
Temperature Products
>
Infrared Thermometers / Sensors
>
Food Infrared Thermometers
>
Oakton Food TempTestr Basic Infrared Thermometer
Share
|
Oakton Food TempTestr Infrared Thermometer
EW-35625-15
$99.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Ideal for food inspections and HACCP programs
Non-contact measurement eliminates the risk of cross-contamination
Choose from IR meter only or IR meter kit
Temperature measurement is essential when assuring proper food storage, cooking, and serving environments. Quick checks with a non-contact thermometer can remove the hazard of cross-contamination and reduce the time a traditional probe requires.
TempTestr IR Food Thermometer
provides safe, non-contact readings in 1⁄2 second. It has a compact design for easy one-handed operation. Other features include a laser sighting to pinpoint your exact target, selectable degF/degC display, steam interference filter, and a hold function that freezes display for 7 seconds after button release. Accuracy of ±2degF (±1degC) over the HACCP critical temperature range of 40 to 140degF (4 to 60 degC).
TempTestr IR Food Thermometer Kit
Specifications
Product Type
Food Infrared Thermometer
Min temperature (º C)
-35
Min temperature (º F)
-31
Max temperature (º F)
572
Max temperature (º C)
275
Voltage input
No
Sighting type
Single-point laser
Distance to target size ratio
4:1
Emissivity
Fixed
Laser class
Class II
Battery life
12 hours
Accuracy (º C)
0 to 65°C: ±1°C
below 0°: ±1°C + 0.1°
above 65°C: 1.5% of reading
Accuracy (º F)
32 to 150°F: ±2°F
below 32°F:
2°F + 0.1°
above 150°F: 1.5% of reading
Repeatability (º C)
within accuracy of unit
Repeatability (º F)
within accuracy of unit
Resolution (º F)
1°F ±2°F at HACCP of critical temperature range
Resolution (º C)
1°C ±1°C at HACCP of critical temperature range
Response time
500 mSec, 95% response
Brand
Oakton
Accessories
Qty
/
Price
Availability
Batteries, 9 V, 4/pack
EW-09376-04
at
$25.60 USD/PACKAGE of 4
In stock.
Add to Cart
Temptestr Beltloop Carrying Case
EW-35625-80
at
$18.50 USD/EACH
In stock.
Add to Cart
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; IR Thermometer
EW-17004-00
at $130.00 USD/EACH
Usually ships in 4 days.
Add to Cart
Technical Resources
Infrared Brochure(1.44MB)
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
IR Temperature Monitors
Basics of Cartridge Filtration
Bitter Brew
Show All...
Temperature FAQs
Temperature Instrument Ranges
Getting a Handle on Temperature
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title