Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Oakton PC2700 Meter w/pH electrode, conductivity/temp probe, and electrode stand

EW-35414-00
$1,300.00 USD/EACH

  • Multiparameter benchtop meters are an all-in-one solution for laboratory research work
  • Advanced research-grade meter
  • Includes Ion concentration readings
Oakton PC 2700 meters feature pH auto-buffer recognition for both USA and NIST buffer sets, automatic or manual temperature compensation, and selectable temperature units. Previous calibration points, electrode slope and offset, and conductivity/TDS cell constants are stored in memory for quick and easy retrieval.

Meters offer an advanced user interface and advanced measurement capabilities. Meters meet GLP requirements—meters stamp all calibration and stored data with time/date. Nonvolatile memory stores up to 500 data sets to simplify documenting large volumes of pH, mV, ISE, conductivity, TDS, salinity, or resistivity measurements. Data is easily exported to a computer via RS-232 port. Order RS-232 and optional RS-232 to USB adapter separately.


What's Included:
universal 100/240 VAC power supply and electrode stand.

Meter with probes adds: combination pH electrode (35805-04) and conductivity/temperature probe (35412-10).

For complete specifications please view the instruction manual
Product Type Multiparameter Meter
Meter style Benchtop
Dimensions (" W) 6-7/8
Dimensions (" H) 2-3/4
Dimensions (" L) 6-1/8
Range (pH) 2.000 to 20.000
Range (mV) ± 2000
Range (Conductivity) 0 to 500 mS
Range (TDS) 0 to 500 ppt
Range (Salinity) 0 to 80 ppt
Range (Resistivity) 0 to 20.00 M ohm
Range (ISE) 0 to 19999 ppm
Range (Temperature °C) 10 to 110
Range (Temperature °F) 14 to 230
Resolution (pH) 0.1/0.01/0.001
Resolution (Temperature) 0.1
Resolution (mV) 0.1/1
Resolution (Conductivity) 0.01, 0.1 µS; 0.001, 0.01, 0.1 mS
Resolution (TDS) 0.01, 0.1 ppm, 0.001, 0.01, 0.1 ppt
Resolution (Resistivity) 0.01 m ohm, 0.001, 0.01 k ohm, 0.01, 0.1 ohm
Resolution (ISE) Up to 3 digits
Resolution (Salinity) 0.01, 0.1 ppm; 0.001, 0.01 ppt
Accuracy (pH) ±0.002 pH
Accuracy (Temperature) ±0.5°C
Accuracy (mV) ±0.2 mV or 0.05%, whichever is greater
Accuracy (Resistivity) ±1% full-scale
Accuracy (ISE) ±0.5% of reading
Accuracy (Conductivity) ±1% full-scale
Accuracy (TDS) ±1% of reading
Accuracy (Salinity) ±1.0% full-scale
Temperature compensation Automatic or manual
Display LCD
Interface RS-232
Calibration Up to 6 points
Connections BNC, Temp 2.5mm
Buffer recognition USA, NIST, DIN
Power 100 to 240 VAC, 50/60 Hz
Datalogging (points) 500
CE Compliance Yes
Brand Oakton

Accessories
Qty/Price
Availability

at $29.00 USD/PACKAGE of 3

In stock.

at $39.00 USD/EACH

In stock.

at $291.00 USD/EACH

In stock.

at $38.50 USD/EACH

In stock.

at $695.00 USD/EACH

In stock.

at $77.00 USD/EACH

In stock.

at $128.00 USD/EACH

In stock.
Calibrations
Qty/Price
Availability

at $115.00 USD/EACH
Usually ships in 9 days.




Technical Resources
 
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title