Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Oakton PC700 Meter and electrode stand

EW-35413-20
$772.00 USD/EACH

  • Easily toggle between pH, mV, conductivity, TDS, and temperature measurements at the touch of a button
  • Space-saving design takes up less room on the bench than other meters
  • Large display lets you see your measurements from across the lab
  • Conductivity and TDS autoranging chooses the best resolution for each measurement
The PC700 meter offers a new compact design that is over 40% smaller than other benchtop meters. The large display is easy to read even at a distance. Dual-display allows you to read both a measured parameter and temperature at a glance. The meter features pH auto-buffer recognition for both USA and NIST buffer sets (1.68, 4.01, 7.00, 10.01 and 12.45 or 1.68, 4.01. 6.18. 9.86, and 12.45), automatic or manual temperature compensation, temperature selectable in either °F or °C, display Hold, and water-resistant membrane keypad.

Previous calibration points, electrode slope and offset, and conductivity/TDS cell constants are stored in memory for quick and easy retrieval. You can also store up to 100 data sets in the meter's memory for retrieval later. The convenient pull-out quick reference card keeps keypad functions at your fingertips.


What's Included:
universal 100/240 VAC power supply, power adapters, and electrode stand.

Meter with probes adds: combination pH electrode (35805-04) and conductivity/temperature probe (35608-74).
Product Type Multiparameter Meter
Meter style Benchtop
Dimensions (" H) 6-7/8
Dimensions (" D) 2-3/4
Dimensions (" L) 6-1/8
Range (pH) 2.00 to 16.00
Range (mV) ± 2000
Range (Conductivity) 0 to 200 mS
Range (TDS) 0 to 200 ppt
Range (Temperature °C) 10 to 110
Range (Temperature °F) 14 to 230
Resolution (pH) 0.1/0.01
Resolution (Temperature) 0.1
Resolution (mV) 0.1/1
Resolution (Conductivity) 0.01, 0.1 µS; 0.001, 0.01, 0.1 mS
Resolution (TDS) 0.01, 0.1 ppm, 0.001, 0.01, 0.1 ppt
Accuracy (pH) ±0.01 pH
Accuracy (Temperature) ±0.5°C
Accuracy (mV) ±0.2 mV or 0.05%, whichever is greater
Accuracy (Conductivity) ±1% full-scale
Accuracy (TDS) ±1% of reading
Temperature compensation Automatic or manual
Display LCD
Calibration Up to 5 points
Connections BNC, Temp 2.5mm
Buffer recognition USA, NIST
Power 100 to 240 VAC, 50/60 Hz
Datalogging (points) 100
CE Compliance Yes
Brand Oakton

Accessories
Qty/Price
Availability

at $29.00 USD/PACKAGE of 3

In stock.

at $77.00 USD/EACH

In stock.

at $131.00 USD/EACH

In stock.

at $84.00 USD/EACH

In stock.
Calibrations
Qty/Price
Availability

at $115.00 USD/EACH
Usually ships in 9 days.




Technical Resources
 
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title