| Product Type |
Multiparameter Meter |
| Meter style |
Benchtop |
| Dimensions (" W) |
6-7/8 |
| Dimensions (" H) |
2-3/4 |
| Dimensions (" L) |
6-1/8 |
| Range (pH) |
2.00 to 16.00 |
| Range (mV) |
± 2000 |
| Range (Conductivity) |
0 to 200 mS |
| Range (TDS) |
0 to 200 ppt |
| Range (Temperature °C) |
10 to 110 |
| Range (Temperature °F) |
14 to 230 |
| Resolution (pH) |
0.1/0.01 |
| Resolution (Temperature) |
0.1 |
| Resolution (mV) |
0.1/1 |
| Resolution (Conductivity) |
0.01, 0.1 µS; 0.001, 0.01, 0.1 mS |
| Resolution (TDS) |
0.01, 0.1 ppm, 0.001, 0.01, 0.1 ppt |
| Accuracy (pH) |
±0.01 pH |
| Accuracy (Temperature) |
±0.5°C |
| Accuracy (mV) |
±0.2 mV or 0.05%, whichever is greater |
| Accuracy (Resistivity) |
±1% full-scale |
| Accuracy (Conductivity) |
±1% full-scale |
| Accuracy (TDS) |
±1% of reading |
| Accuracy (Salinity) |
±1.0% full-scale |
| Temperature compensation |
Automatic or manual |
| Display |
LCD |
| Calibration |
Up to 6 points |
| Connections |
BNC, Temp 2.5mm |
| Buffer recognition |
USA, NIST, DIN |
| Power |
100 to 240 VAC, 50/60 Hz |
| Datalogging (points) |
100 |
| CE Compliance |
Yes |
| Brand |
Oakton |