dummy
EW-35425-10
- The ultimate tester for pH, conductivity, TDS, salinity and temperature!
- Combine measurement capabilities of three testers in one convenient pocket-sized unit
- Up to 5 point pH calibration with NIST or USA autobuffer recognition
- Conductivty, TDS, and salinity feature autoranging based on the concentration of your sample
- IP67 - rated housing floats if accidentally dropped in water
Great for water, wastewater, laboratory or plant use, these multiparameter Testrs feature long-life pH electrodes with a wide range of sample compatibility. Pin-style conductivity sensors feature stainless steel electrodes for chemical resistance and durability. Replaceable sensor module saves you money by reusing the meter body. Advanced features include auto-shutoff, user-adjustable temperature coefficient, self-adjusting TDS and salinity factor, and low-battery indicator.
What's Included:
protective plastic storage case, lanyard, and batteries.
| Product Type |
pH Tester |
| Meter style |
Tester |
| Dimensions (" L) |
6-1/2 |
| Dimensions (" Dia) |
1-1/2 |
| Range (pH) |
0.00 to 14.00 |
| Range (Conductivity) |
0 to 20.0 mS |
| Range (TDS) |
0 to 10.0 ppt |
| Range (Salinity) |
0 to 10 ppt |
| Range (Temperature °C) |
10 to 110 |
| Range (Temperature °F) |
14 to 230 |
| Resolution (pH) |
0.1/0.01 |
| Resolution (Temperature) |
0.1 |
| Resolution (Conductivity) |
0.1, 1 µS; 0.01 mS |
| Resolution (TDS) |
0.01, 0.1 ppm, 0.001, 0.01, 0.1 ppt |
| Resolution (Salinity) |
0.1 ppt, 0.01% |
| Accuracy (pH) |
±0.01 pH |
| Accuracy (Temperature) |
±0.5°C |
| Accuracy (Conductivity) |
±1% full-scale |
| Accuracy (TDS) |
±1% of reading |
| Accuracy (Salinity) |
±1.0% full-scale |
| Temperature compensation |
Automatic |
| Display |
LCD |
| Calibration |
Up to 5 points |
| Waterproof |
Yes |
| Buffer recognition |
USA, NIST |
| Power |
Four AA batteries (included) |
| Battery life |
150 hours |
| CE Compliance |
Yes |
| Brand |
Oakton |
| Model |
PCSTestr 35 |
at $29.00 USD/PACKAGE of 3
In stock.
at $15.50 USD/PACKAGE of 6
In stock.
at $74.00 USD/EACH
In stock.
at $115.00 USD/EACH
Usually ships in 9 days.
Technical Resources
Related technical articles (Roll over for summary)
Show Less...
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334