Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Home  > 
Discontinued Items >
Oakton RH/TempLog Temperature/ Humidity Datalogger
Item# EW-35710-10
$192.00 USD/EACH
Discontinued
Qty
  • Compact, rugged housing guarantees operation in busy or rugged areas!
  • Sample from once per 10 seconds to once per 2 hours—built-in clock and calendar track your data in real time
  • Quickly document min/max values with optional IR printer
  • Built-in clock and calendar track your data in real time
These loggers store up to 16,000 measurements, allowing you to track important temperature and humidity data for months. The two-digit LCD shows current conditions—minimum and maximum readings are available at the touch of a button. If set limits are passed, the built-in alarm flickers the display and must be manually reset. Use the external sensor input to add one additional sensor (sold separately) to your datalogger.
Model 35710-00 records temperature and features a water-resistant IP65-rated housing. Model 35710-10 records temperature and humidity and features a dustproof IP60-rated housing. 35710-60 and 35710-62 Data Logger kits include datalogger, software and cable.
35710-50 software/cable (sold separately) is required for use with the 35710-10 Data Logger
Product Type Temperature/Humidity Dataloggers
RH accuracy ±3%RH from 0 to 90% RH
±6%RH from 90 to 100% RH
Temp resolution 1.0°F(0.5°C)
RH range 0 to 100% RH
Accuracy ±3%RH (0 to 90%RH), ±6%RH (90 to 100%RH)
Display Two digit LCD display, 0.375 in H
No. of channels 1 internal, 1 external
Battery life Approx. 2 years
Memory 16,000 data points
Model EC650
Manufacturer number EC650
Power 3.6 V lithium battery
Brand Oakton
Accessories
Qty/Price
Availability
at $385.00 USD/EACH
Usually ships in 11 days.
Calibrations
Qty/Price
Availability
at $176.00 USD/EACH
Usually ships in 9 days.
Technical Resources
Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
Questions and Answers
Do you have questions about this product?
Get answers from real customers and in-house experts with AnswerBox.
3 Questions | 4 Answers
Displaying questions 1-3
  • Q:
    The tech specs show that the cable requires a serial port. This seems very antiquated. Is there a USB cable or a wireless option?
    Asked on 12/28/2012 by Wendy
    1 answer
    • STAFF EXPERT
      A:
      You can use the IRDA (infrared data assembly) interface to dowload data to the printer, catalog number 92600-40. IRDA is a wireless technology.
      Answered on 12/28/2012 by Christina from Technical Support
  • Q:
    The unit is described as durable, but is it water proof? Could it be utilized in an extremely warm, extremely wet environment like a salt fog chamber?
    Asked on 9/14/2010 by Jackie from Rockford, IL
    1 answer
    • STAFF EXPERT
      A:
      The casing of this product is rated IP60, which is total dust ingress protection, and low moisture protection. I think a different product would be a better fit for a salt fog chamber. Please contact our technical application specialists for additional assistance (contact information can be found in the "contact us" section on the web page).
      Answered on 9/16/2010 by Jack from Technical Support
  • Q:
    We presently use a hygro thermograph equipment (weekly charts) to record RH/Temp in a laboratory. However we would like to change to a digital recorder for convenience and to be able to store the information in a digital format rather than storing used charts. Would this equipment suit our needs.
    Asked on 9/7/2009 by Hernan from Belize. C.A
    2 answers
    • STAFF EXPERT
      A:
      Yes, this unit would work well for your application, please note that loggers require 35710-50 software/cable if not used with 35710-20 cradle & 35710-25 receiver. Please contact our technical application specialists for additional assistance (contact information can be found on the product web page).
      Answered on 9/9/2009 by Jen from Technical Support
    • VERIFIED BUYER
      A:
      If +/- 1 Deg F and +/- 3% RH is good enough, these might be a great solution.
      If you add a cradle to each datalogger and buy a receiver antenna you can setup wireless data monitoring and collection as well, as long as you have near line-of-sight with each datalogger. That would save the need to retrieve each datalogger for your weekly downloads.
      Answered on 6/6/2010 by TT from Seattle, WA
Displaying questions 1-3
Do you have a question about this product? 
Product Reviews
REVIEW SNAPSHOT®
by PowerReviews
Oakton<reg> RH/TempLog Temperature/ Humidity Datalogger
3.7
(based on 6 reviews)
80%
of respondents would recommend this to a friend.
Pros
  • Durable (3)
  • Easy to use (3)
Cons
No Cons
Best Uses
  • In the field (4)
Reviewed by 6 customers
Sort by
Displaying reviews 1-6
Back to top
(1 of 1 customers found this review helpful)
3.0
Oakton RH Museum Use
By Archivist
from Ontario, Canada
Verified Reviewer
Pros
    Cons
      Best Uses
      • In The Field
      • Research
      Comments about Oakton<reg> RH/TempLog Temperature/ Humidity Datalogger:
      I work in a museum and we are using 14 of these loggers at the present time. They are easy to set up and to read, are probably good for temporary or short term uses, but I am not sure I would recommend them for a museum.
      For one, they are not Mac compatible. This means the purchase of an extra computer specifically for this purpose, and importing and exporting to make the files transferrable to a Mac.
      I have also just discovered that when the battery gets low, they stop recording. They do not show any indication that the battery is low, and the display still works, so you can't tell they are not recording until you download the information onto the computer. I have lost 3 weeks worth of recordings from the last month due to this issue. Some sort of low battery indicator on the front of the unit would be good.
      4.0
      Used one, now we use 3.
      By Boom
      from Minden, LA
      Verified Buyer
      Pros
      • Accurate
      • Calib within- 2F - 2
      • Durable
      • Easy To Use
      • Works Properly
      Cons
        Best Uses
        • In The Field
        Comments about Oakton<reg> RH/TempLog Temperature/ Humidity Datalogger:
        Came calibrated +/- 2F, +/-2%, the data logger works well in the environment that we have inside our plant. I do wish that they would update the computer cable to a flat USB plug instead of the RS.
        4.0
        Rugged
        By TT
        from Seattle, WA
        Verified Buyer
        Pros
        • Durable
        • Easy To Use
        Cons
          Best Uses
          • In The Field
          Comments about Oakton<reg> RH/TempLog Temperature/ Humidity Datalogger:
          I take these with me to industrial sites for use in performance assessments where equipment is sensitive to ambient conditions. They are easy to setup and install. [...] conditions I've put them trough - with one exception - a location with high winds and super-saturated air. Condensation on the datalogger sensors did cause it to fail - temporarily. Once they dried out, they started working again and passed calibration with flying colors.I would like something with higher accuracy, but these are so easy to use, they're hard to beat.
          2.0
          Recent Purchase Does Not Work
          By only half work
          from Valencia, CA
          Pros
            Cons
            • Doesn't Work Properly
            Best Uses
            • In The Lab
            • Research
            Comments about Oakton<reg> RH/TempLog Temperature/ Humidity Datalogger:
            I own 6 wireless units for temperature readings. The 1st three I bought work just fine, still do. The next three I purchased (NIST calibrated mind you) cannot even sync to the "home base" computer. Original work without a hitch. Next three will not sync, transmit, or record. [...] If the rating were on the last 3 I purchased I'd give it a zero, but since I own 3 that DO work I give it a two.
            5.0
            Oakton RH/T datalogger performance
            By Susana
            from Lima - Peru
            About Me Academic/Professional Researcher
            Pros
            • Accurate
            • Durable
            • Easy To Use
            • Well Calibrated
            • Works Properly
            Cons
              Best Uses
              • In The Field
              • In The Lab
              Comments about Oakton<reg> RH/TempLog Temperature/ Humidity Datalogger:
              I used it to register T and RH outdoors and indoors. It is very easy to use and strong. I recomnmend it.
              The only thing not so good is the battery model it uses, that can be difficult to find.
              (1 of 1 customers found this review helpful)
              4.0
              Oakton RH/Temperature Loggers
              By QC Brad
              from York, PA.
              Verified Buyer
              Pros
              • High Quality
              Cons
              • Expensive
              Best Uses
                Comments about Oakton<reg> RH/TempLog Temperature/ Humidity Datalogger:
                The data loggers are used at our casket manufacturing facility to capture both moisture and temperature readings. The data is then downloaded onto a spreadsheet and the Equilibrium Moisture Content (EMC) determed for that particular time frame. Determining variations in EMC by Department enables Quality Control to determine if moisture values are relevant to production scrap with regard to wood defects.
                Displaying reviews 1-6
                Back to top
                Customers Who Bought This Item Also Bought