United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
Representative photo only
Oakton TDSTestr 11 dual range pin style pocket TDS tester
Click to enlarge
More Product Details
Specifications
Accessories
Calibrations
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Related media
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-35662-10
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17106-22
NIST-Traceable calibration for conductivity pens
$65.00 USD/EACH/EACH
Usually ships in 8 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
pH Meters and Water Quality Products
>
Pocket Testers
>
Oakton TDS Pocket Testers
>
Oakton TDSTestr 11 Dual-Range Pocket TDS Testers
Share
|
Oakton TDSTestr 11 dual-range, pin-style pocket TDS tester
EW-35662-10
$83.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Dual-range measurements increase application range
Replaceable sensor prolongs the life of your instrument
Single or one point per range calibration
Auto shutoff, low-battery indicator, error messages
Never watch a tester sink to the bottom of your tank or pond! These waterproof testers float, so there's no need to worry. Valox
®
housing and stainless steel electrodes offer good chemical compatibility and durability. Instrument features a replaceable sensor. Reuse the meter body many times without losing waterproof and dustproof protection.
What's Included:
protective plastic storage case, lanyard, and batteries.
Specifications
Product Type
TDS Tester
Meter style
Tester
Dimensions (" W)
6-1/2
Dimensions (" Dia)
1-1/2
Range (TDS)
0 to 10.0 ppt
Range (Temperature °C)
5 to 50
Range (Temperature °F)
23 to 221
Resolution (Temperature)
0.1
Resolution (TDS)
0.01, 0.1 ppm, 0.001, 0.01, 0.1 ppt
Accuracy (Temperature)
±0.5°C
Accuracy (TDS)
±1% of reading
Temperature compensation
Automatic
Display
LCD
Calibration
Up to 3 points
Waterproof
Yes
Power
Four 1.5 V batteries (included)
Battery life
150 hours
CE Compliance
Yes
Brand
Oakton
Model
TDSTestr 11
Accessories
Qty
/
Price
Availability
Batteries, 1.5V button cells, 6/PK
EW-09377-16
at
$15.50 USD/PACKAGE of 6
In stock.
Add to Cart
Soft carrying case for waterproof testers
EW-35624-45
at
$27.80 USD/EACH
In stock.
Add to Cart
Replacement sensor for Oakton TDS, EC, and Salt Testrs
EW-35661-17
at
$50.00 USD/EACH
In stock.
Add to Cart
Calibration kit for Oakton standard and dual-range TDSTestrs
EW-35661-70
at
$41.50 USD/EACH
In stock.
Add to Cart
Calibrations
Qty
/
Price
Availability
NIST-Traceable calibration for conductivity pens
EW-17106-22
at $65.00 USD/EACH
Usually ships in 8 days.
Add to Cart
Technical Resources
Instruction Manual
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
As Easy as T-D-S!
Conductivity FAQs
Conductivity Theory and Technical Tips
Show All...
Understanding Low Conductivity/High Resistivity Measurements
Temperature Effects on Conductivity Measurement
Conductivity Technical Information
Testing the pH of Wastewater Samples
Testing the pH of Soil Samples
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Related media
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title