dummy
EW-35635-00
- Waterproof and dustproof with IP67-rated housing that floats!
- Meets performance criteria as specified by ISO 7027 (DIN EN 27027) method
- Compliant with ASTM D6855-03 (EPA recognized)
- Four point push-button calibration for full-range accuracy
- Auto-ranging from 0 to 1000 NTU
The Oakton T-100 turbidity meter is easy to calibratemeter automatically prompts you to the next calibration standard. The large custom LCD displays reading and units of measure. Other features include auto-off, diagnostic error messages, and auto-ranging function. Power saving function lets you take over 1000 readings on a single set of AAA batteries. Sealed optical system with infrared light source delivers high accuracy across entire range.
Meter also includes 60-mL of four primary standards (0.02, 20.0, 100 and 800 NTU) in HPTE bottles,with four labeled cuvettes for standards.
What's Included:
three samlple cuvettes, a light shield cap, 10-mL silicone oil, lint-free cloth, 4 AAA batteries, and hard carrying case.
| Product Type |
Turbidity Meter |
| Meter style |
Portable |
| Dimensions (" W) |
2-3/4 |
| Dimensions (" H) |
2 |
| Dimensions (" L) |
6-1/2 |
| Range (Turbidity) |
0.01 to 1000 |
| Range (Temperature °C) |
0 to 50 |
| Range (Temperature °F) |
32 to 122 |
| Resolution (Turbidity) |
0.01, 0.1, 1 |
| Accuracy (Turbidity) |
± 2% of reading |
| Display |
LCD |
| Waterproof |
Yes |
| Power |
Four AAA batteries (included) |
| CE Compliance |
Yes |
| Brand |
Oakton |
at $20.04 USD/EACH
Call 1-888-358-4705 for availability.
at $21.25 USD/PACKAGE of 12
In stock.
at $297.00 USD/EACH
In stock.
at $40.00 USD/PACKAGE
In stock.
Technical Resources
Related technical articles (Roll over for summary)
Show All...
Show Less...
Still have questions?
Contact Our Experts
for FREE product
application selection