log in or register
Oakton Waterproof DO 300 Meter with Probe
EW-35641-00
$744.00 USD/EACH
  • IP67 waterproof and dustproof housings float
  • Perfect for harsh, rugged field conditions
  • Store 50 measurements with corresponding temperatures and time
  • Galvanic probe allows you to take measurements immediately
Dissolved oxygen (DO) readings are displayed in mg/L, ppm, or % saturation simultaneously with temperature values. Adjustable backlit display makes it easy to view readings in low light levels. Calibrate at the independent 100% and 0% calibration points to achieve high accuracy over the entire range. Increase reading accuracy by entering barometric pressure and salinity—meters automatically calculate offset. Setup function displays electrode slope, zero offset, and millivolt. Real-time clock stamps stored readings and calibration data with date and time, making it ideal for Good Laboratory Practice (GLP) standards. Additional features include selectable °C or °F temperature units, Hold function, ready indicator, and auto-off for battery conservation.
What's Included:
batteries.
Meter with probe adds: dissolved oxygen probe, replacement membrane cap, and electrolyte solution.
Product Type Dissolved Oxygen Meter
Meter style Portable
Dimensions (" W) 3-1/2
Dimensions (" H) 1-3/4
Dimensions (" L) 7-1/2
Range (Dissolved oxygen) 0 to 19.99mg/L
Range (Temperature °C) 5 to 50
Range (Temperature °F) 23 to 221
Resolution (Temperature) 0.1
Accuracy (Temperature) ±0.3°C
Accuracy (Dissolved Oxygen) ±1.5% full scale
Temperature compensation Automatic
Display LCD
Waterproof Yes
Power Four AAA batteries (included)
Datalogging (points) 50
CE Compliance Yes
Brand Oakton
Accessories
Qty/Price
Availability
at $16.50 USD/PINT
In stock.
at $21.25 USD/PACKAGE of 12
In stock.
at $66.00 USD/EACH
In stock.
at $415.00 USD/EACH
In stock.
at $463.00 USD/EACH
In stock.
at $555.00 USD/EACH
Call 1-888-358-4705 for availability.
at $711.00 USD/EACH
Call 1-888-358-4705 for availability.
at $101.00 USD/EACH
In stock.
at $21.50 USD/EACH
In stock.
at $64.00 USD/EACH
In stock.
Calibrations
Qty/Price
Availability
at $115.00 USD/EACH
Usually ships in 16 days.
Technical Resources
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title