dummy
EW-35423-10
- Easy-to-read display with an easy-to-afford price!
- Ergonomically designed LCD means measurements can be seen at a glance
- Economical and easy to use
- Waterproof and dustproof housing designed for use in dirty and damp field conditions
Simply dip and read the orientation of the display means no need to turn your head or tilt the tester. This low-cost meter is perfect for most applications. The single-junction electrode is recessed to protect against breakage. The unit offers up to three-point calibration with auto-buffer recognition for quick calibrations, automatic temperature compensation for accurate reading under changing conditions, hold function to lock measure value, and auto-off to prolong battery life. Built-in belt clip allows you to hang your meter when not in use.
What's Included:
four 1.5 V button cell batteries.
| Product Type |
pH Meters |
| Meter style |
Portable |
| Dimensions (" W) |
1-5/8 |
| Dimensions (" H) |
6-1/4 |
| Dimensions (" D) |
1-1/8 |
| Range (pH) |
0.0 to 14.0 |
| Range (Temperature °C) |
0 to 50 |
| Range (Temperature °F) |
32 to 122 |
| Resolution (pH) |
0.1 |
| Accuracy (pH) |
±0.1 |
| Temperature compensation |
Automatic |
| Display |
LCD |
| Calibration |
3 |
| Waterproof |
Yes |
| Buffer recognition |
USA, NIST |
| Power |
Four 1.5 V button cell batteries (included) |
| Brand |
Oakton |
| Model |
EcoTestr pH 2 |
at $29.00 USD/PACKAGE of 3
In stock.
at $15.50 USD/PACKAGE of 6
In stock.
at $42.00 USD/EACH
In stock.
at $31.90 USD/PACKAGE of 20
Call 1-888-358-4705 for availability.
at $65.00 USD/EACH
Usually ships in 9 days.
Technical Resources
Virtual Catalog (PDFs)
More info
Related technical articles (Roll over for summary)
Show All...
Show Less...
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334