United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
EW-35650-10
Oakton
®
Waterproof ORPTestr
®
10 Pocket ORP Tester
Click to enlarge
More Product Details
Specifications
Accessories
Calibrations
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
More info
Brochure
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-35650-10
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17101-45
NIST Traceable Calibration; pH Tester and Pens
$65.00 USD/EACH/EACH
Usually ships in 9 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
pH Meters and Water Quality Products
>
pH Meters, Ion Meters and ISFET Meters
>
pH/Ion Pocket Testers
>
Oakton Waterproof ORPTestr 10 Pocket ORP Tester
Share
|
Oakton Waterproof ORPTestr 10 Pocket ORP Tester
EW-35650-10
$194.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Ideal for checking chromate reduction, cyanide oxidation, and other ORP readings
Large, horizontal readout
IP67-rated housing floats in water and sits flat on a hard surface
ORPTestr 10 features a replaceable double-junction platinum electrodeideal for checking dirty water, heavy metals, sulfides, and organics. Valox
®
electrode body resists most chemicals.
Use these versatile instruments for applications ranging from swimming pool testing to pulp bleaching. Meter features ±2 mV accuracy and a ±150 mV offset. Housing allows you to attach a lanyard (included) for easy retrieval. Additional features include a hold function, auto-off function, self-diagnostic error messages, and a three-segment battery-life indicator.
Specifications
Product Type
ORP Tester
Meter style
Tester
Dimensions (" L)
6
Dimensions (" Dia)
1-1/2
Range (mV)
±1000
Range (Temperature °C)
5 to 50
Range (Temperature °F)
23 to 221
Resolution (mV)
0.1/1
Accuracy (mV)
±2 mV
Display
LCD
Waterproof
Yes
Power
Four 1.5 V batteries (included)
CE Compliance
Yes
Brand
Oakton
Model
ORPTestr® 10
Accessories
Qty
/
Price
Availability
YSI Zobell solution, 1.25 g
EW-05478-60
at
$31.00 USD/EACH
In stock.
Add to Cart
Batteries, 1.5V button cells, 6/PK
EW-09377-16
at
$15.50 USD/PACKAGE of 6
In stock.
Add to Cart
Soft carrying case for waterproof testers
EW-35624-45
at
$27.80 USD/EACH
In stock.
Add to Cart
ORPTestr
replacement sensor
EW-35650-09
at
$91.00 USD/EACH
In stock.
Add to Cart
Calibrations
Qty
/
Price
Availability
NIST Traceable Calibration; pH Tester and Pens
EW-17101-45
at $65.00 USD/EACH
Usually ships in 9 days.
Add to Cart
Technical Resources
Instruction Manual (118 KB)
Oakton pH Testrs (903 KB)
Virtual Catalog (PDFs)
13-14 Catalog
More info
Oakton Catalogo en Espanol.pdf (8.3 MB)
Related technical articles (Roll over for summary)
Double Junction pH Tester-The Next Generation of Long Life Pocket-Sized pH Meters
pH FAQs
Testing the pH Value of Pure Water
Show All...
Testing the pH value of Meat
Testing the pH in Yogurt Production
Testing the pH value of Wine
Testing the pH of Wastewater Samples
Testing the pH of Soil Samples
pH Electrode Theory
Oakton® pH meter becomes a TV star!
Measuring the pH of Pure Water
pH Systems
pH Temperature Compensation
Temperature Compensation for pH Instruments
pH Measurement FAQs
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title