Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Oakton Waterproof pHTestr 10 BNC Pocket pH Tester

EW-35634-14
$108.00 USD/EACH

  • Customizable to your application—use with any pH electrode with a BNC connector
  • Versatility of a handheld meter at a pocket price
  • IP67-rated protects meter from water, dust, and shock while offering chemical resistance
  • Meter floats—accidental drops into water will not result in lost or damaged meters
Simply choose the electrode best suited for your application, connect the pHTestr BNC 10, calibrate, and measure. Change electrodes to suit your application without having to purchase another meter. Order electrodes separately.

Large display and tactile keypad allow for easy calibration along with USA and NIST auto-buffer recognition. Hold function freezes reading on the display for easy recording. Additional features include Valox® sensor housing, self-diagnostic error messages, a battery-life indicator, and an auto-off function to maximize battery life.
What's Included:
protective plastic storage case, lanyard, and batteries.
Product Type pH Tester
Meter style Tester
Dimensions (" L) 6
Dimensions (" Dia) 1-1/2
Range (pH) 0.00 to 14.00
Range (Temperature °C) 5 to 50
Range (Temperature °F) 23 to 221
Resolution (pH) 0.1/0.01
Accuracy (pH) ±0.01 pH
Temperature compensation Manual
Display LCD
Calibration Up to 3 points
Connections BNC, Temp 8-pin mini-DIN
Waterproof Yes
Buffer recognition USA, NIST
Power Four 1.5 V batteries (included)
CE Compliance Yes
Brand Oakton
Model pHTestr® 10 BNC

Accessories
Qty/Price
Availability

at $29.00 USD/PACKAGE of 3

In stock.

at $15.50 USD/PACKAGE of 6

In stock.

at $27.80 USD/EACH

In stock.

at $42.00 USD/EACH

In stock.

at $45.90 USD/EACH

In stock.

at $52.02 USD/EACH

In stock.

at $67.32 USD/EACH

In stock.
Related products
Qty/Price
Availability

at $108.00 USD/EACH
In stock.
Calibrations
Qty/Price
Availability

at $115.00 USD/EACH
Usually ships in 9 days.




Technical Resources
 
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title