dummy
EW-35634-20
- Versatile, compact design works in a field or laboratory setting
- pHTestr 20 features ±0.01 pH accuracysuitable for most ASTM and EPA test procedures.
- A complete meter with key features at an economical price
- Replaceable, double-junction electrode sensors save time and money
- IP67-rated housing sits flat on the table and floats in water
Waterproof pHTestr 20 is great for outdoor applications such as taking pH measurements in lakes and streams. Designed to float in water, you can even attach a line to the housing for easy retrieval. Valox® sensor housing resists most chemicals.
The pHTestr 20 has a replaceable sensor for easy maintenance. Double-junction electrode sensor handles wastewater and solutions containing sulfides, heavy metals, and TRIS. Electrode sensor offers a clog-resistant Kynar® junction and a recessed, glass pH bulb to protect against breakage.
The keypad clicks to confirm operation, and allows access to a variety of functions. Calibrate up to three pH points with the auto-buffer recognition at both USA and NIST standard buffers. Additional features include a automatic temperature compensation (ATC), hold function, auto-off function, self-diagnostic error messages, and a three-segment battery-life indicator.
Meter includes: protective plastic storage case, lanyard, and batteries.
Meter kit adds: two single-use pouches of pH 4.01, pH 7.00, pH 10.0 buffer, and rinse solution, one sample jar, and sturdy carrying case.
| Product Type |
pH Tester |
| Meter style |
Tester |
| Dimensions (" L) |
6-1/2 |
| Dimensions (" Dia) |
1-1/2 |
| Range (pH) |
0.00 to 14.00 |
| Range (Temperature °C) |
5 to 50 |
| Range (Temperature °F) |
23 to 221 |
| Resolution (pH) |
0.1/0.01 |
| Accuracy (pH) |
±0.01 pH |
| Temperature compensation |
Automatic |
| Display |
LCD |
| Calibration |
Up to 3 points |
| Waterproof |
Yes |
| Buffer recognition |
USA, NIST |
| Power |
Four 1.5 V batteries (included) |
| CE Compliance |
Yes |
| Brand |
Oakton |
| Model |
pHTestr 20 |
at $29.00 USD/PACKAGE of 3
In stock.
at $15.50 USD/PACKAGE of 6
In stock.
at $70.00 USD/EACH
In stock.
at $27.80 USD/EACH
In stock.
at $42.00 USD/EACH
In stock.
at $65.00 USD/EACH
Usually ships in 9 days.
Technical Resources
Related technical articles (Roll over for summary)
Show All...
Show Less...
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334