United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
2 >
EW-35634-30
Oakton Waterproof pHTestr 30 Pocket pH Tester
Click to enlarge
More Product Details
Specifications
Accessories
Calibrations
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
Brochure
Related technical articles
Service & Support
Related media
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-35634-30
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17101-45
NIST Traceable Calibration; pH Tester and Pens
$65.00 USD/EACH/EACH
Usually ships in 9 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
pH Meters and Water Quality Products
>
Pocket Testers
>
Oakton Waterproof pH Testr Pocket pH Testers
>
Oakton Waterproof pHTestr 30 Pocket pH Tester
Share
|
Oakton Waterproof pHTestr 30 Pocket pH Tester
EW-35634-30
$116.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Versatile, compact design works in a field or laboratory setting
±0.01 pH accuracy along with dual pH and selectable °C or °F temperature display
A complete meter with key features at an economical price
Replaceable, double-junction electrode sensors save time and money
IP67-rated housing sits flat on the table and floats in water
Waterproof pHTestr 30 is great for outdoor applications such as taking pH measurements in lakes and streams. Designed to float in water, you can even attach a line to the housing for easy retrieval. Valox
®
sensor housing resists most chemicals.
The pHTestr 30 has a replaceable sensor for easy maintenance. Double-junction electrode sensor handles wastewater and solutions containing sulfides, heavy metals, and TRIS. Electrode sensor offers a clog-resistant Kynar
®
junction and a recessed, glass pH bulb to protect against breakage.
The keypad clicks to confirm operation, and allows access to a variety of functions. Calibrate up to three pH points with the auto-buffer recognition at both USA and NIST standard buffers. Additional features include a automatic temperature compensation (ATC), hold function, auto-off function, self-diagnostic error messages, and a three-segment battery-life indicator.
What's Included:
Meter includes: protective plastic storage case, lanyard, and batteries.
Meter kit adds: two single-use pouches of pH 4.01, pH 7.00, pH 10.0 buffer, and rinse solution, one sample jar, and sturdy carrying case.
Specifications
Meter style
Tester
Dimensions (" L)
6 1/2
Dimensions (" Dia)
1 1/2
Range (pH)
-1 to 15 pH
Range (Temperature °C)
0 to 50
Range (Temperature °F)
32 to 122
Resolution (pH)
0.01
Accuracy (pH)
+/- 0.01
Temperature compensation
Automatic
Display
LCD
Calibration
Up to 3
Waterproof
Yes
Buffer recognition
USA, NIST
Power
Four 1.5V batteries
Battery life
500 hours
CE Compliance
Yes
Accessories
Qty
/
Price
Availability
Oakton buffer pack, 500-ml each of 4.01, 7.00, and 10.00 buffer
EW-05942-10
at
$29.00 USD/PACKAGE of 3
In stock.
Add to Cart
Batteries, 1.5V button cells, 6/PK
EW-09377-16
at
$15.50 USD/PACKAGE of 6
In stock.
Add to Cart
Replacement double-junction sensor for Oakton waterproof pH Testrs
3, 3+, 10, 20, 30
EW-35624-38
at
$70.00 USD/EACH
In stock.
Add to Cart
Soft carrying case for waterproof testers
EW-35624-45
at
$27.80 USD/EACH
In stock.
Add to Cart
Deluxe calibration kit
EW-35624-70
at
$42.00 USD/EACH
In stock.
Add to Cart
Calibrations
Qty
/
Price
Availability
NIST Traceable Calibration; pH Tester and Pens
EW-17101-45
at $65.00 USD/EACH
Usually ships in 9 days.
Add to Cart
Technical Resources
Instruction Manual
Oakton Catalogo en Espanol.pdf (8.3 MB)
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Le secret pour assurer le bon fonctionnement d'un pH-mètre? Les électrodes!
The Secret to Well-Functioning pH Meters? (It's the Electrodes!)
Oakton pHTestr Sensor Module Replacement
Show All...
Double Junction pH Tester-The Next Generation of Long Life Pocket-Sized pH Meters
pH FAQs
Testing the pH Value of Pure Water
Testing the pH value of Meat
Testing the pH in Yogurt Production
Testing the pH value of Wine
Testing the pH of Wastewater Samples
Testing the pH of Soil Samples
pH Electrode Theory
Oakton® pH meter becomes a TV star!
Measuring the pH of Pure Water
pH Systems
pH Temperature Compensation
Temperature Compensation for pH Instruments
pH Measurement FAQs
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Related media
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title