Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Oakton Waterproof pH/Con 10 Meter with pH/conductivity probe

EW-35630-02
$744.00 USD/EACH

  • No more headaches with multiple probes--one probe does it all!
  • Toggle between conductivity and pH with a button press.
  • Features automatic buffer recognition, auto-off, HOLD, and water-resistant membrane keypad
Three-in-one multi-sensor probe with 10-ft (3-m) submersible cable measures pH, conductivity, and temperature. Three-point pH calibration and four-point conductivity calibration provide high accuracy.


What's Included:
batteries and instructions.

Meter with probe adds: pH/conductivity probe 35630-50.

Meter kit further adds: calibration solution pouches (three each of pH 4.01, 7.00, 10.00 and two each of conductivity 447 µS, 1413 µS, 2764 µS, 15,000 µS), three rinse water pouches, sample bottle, instructions, and hard carrying case.
Product Type Multiparameter Meter
Meter style Portable
Dimensions (" W) 3-1/2
Dimensions (" H) 7-3/8
Dimensions (" D) 1-3/4
Range (pH) 0.00 to 14.00
Range (Conductivity) 0 to 19.90 mS
Range (Temperature °C) 10 to 110
Range (Temperature °F) 14 to 230
Resolution (pH) 0.1/0.01
Resolution (Temperature) 0.1
Resolution (Conductivity) 0.01, 0.1 µS; 0.001, 0.01, 0.1 mS
Accuracy (pH) ±0.01 pH
Accuracy (Temperature) ±0.5°C
Accuracy (Conductivity) ±1% full-scale
Temperature compensation Automatic
Display LCD
Calibration Up to 3 points
Waterproof Yes
Buffer recognition USA, NIST
Power Four AAA batteries (included)
Battery life >100 hours
CE Compliance Yes
Brand Oakton

Accessories
Qty/Price
Availability

at $29.00 USD/PACKAGE of 3

In stock.

at $209.00 USD/EACH

In stock.

at $209.00 USD/EACH

In stock.

at $227.00 USD/EACH

In stock.

at $304.00 USD/EACH

Call 1-888-358-4705 for availability.
pH Con Accessory Kit
EW-35630-69

at $87.00 USD/EACH

In stock.
Calibrations
Qty/Price
Availability

at $115.00 USD/EACH
Usually ships in 9 days.




Technical Resources
 
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title