Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Oakton Waterproof pH/Con 300 Meter with probes

EW-35631-00
$821.00 USD/EACH

  • Separate pH and conductivity/TDS probes allow for greater flexibility
  • IP67-rated pH/CON 300 meter floats!
  • Dual display of pH, conductivity or TDS plus temperature (°F/°C selectable)
  • Perform five-point pH and conductivity (TDS) calibration for better accuracy
Features include automatic pH buffer recognition, auto ranging for conductivity/TDS measurements, auto-off, HOLD, and selectable temperature (°C or °F). Meter recalls previous calibration data and pH electrode slope and offset.


What's Included:
batteries and instruction manual.

Meter with probe adds: double-junction combination pH electrode 59001-70, conductivity/temperature probe 35608-50, and four AAA batteries.

Meter kit includes: single-junction pH electrode 59001-65, conductivity/temperature probe 35608-50, calibration solution pouches (three each of pH 4.01, 7.00, 10.00 and two each of conductivity 447 µS, 1413 µS, 2764 µS, 15,000 µS), three rinse water pouches, sample bottle, batteries, and hard carrying case.
Product Type Multiparameter Meter
Meter style Portable
Dimensions (" H) 3-1/2
Dimensions (" D) 1-3/4
Dimensions (" L) 7-1/2
Range (pH) 2.00 to 16.00
Range (Conductivity) 0 to 199.9 mS
Range (TDS) 0 to 200 ppt
Range (Temperature °C) 10 to 110
Range (Temperature °F) 14 to 230
Resolution (pH) 0.1/0.01
Resolution (Temperature) 0.1
Resolution (Conductivity) 0.01, 0.1 µS; 0.001, 0.01, 0.1 mS
Resolution (TDS) 0.01, 0.1 ppm, 0.001, 0.01, 0.1 ppt
Accuracy (pH) ±0.01 pH
Accuracy (Temperature) ±0.5°C
Accuracy (Conductivity) ±1% full-scale
Accuracy (TDS) ±1% of reading
Temperature compensation Automatic or manual
Display LCD
Calibration Up to 5 points
Waterproof Yes
Buffer recognition USA, NIST
Power Four AAA batteries (included)
CE Compliance Yes

Accessories
Qty/Price
Availability

at $29.00 USD/PACKAGE of 3

In stock.

at $129.00 USD/EACH

In stock.

at $157.00 USD/EACH

Call 1-888-358-4705 for availability.
pH Con Accessory Kit
EW-35630-69

at $87.00 USD/EACH

In stock.

at $131.00 USD/EACH

In stock.

at $84.00 USD/EACH

In stock.
Calibrations
Qty/Price
Availability

at $115.00 USD/EACH
Usually ships in 9 days.




Technical Resources
 
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title