United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 5
of
6 >
EW-35614-80
Oakton
®
pH 11 meter kit
Click to enlarge
More Product Details
Specifications
Accessories
Calibrations
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
Brochure
Related technical articles
Service & Support
Related media
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-35614-80
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17106-20
NIST-Traceable Calibration; pH Meter
$115.00 USD/EACH/EACH
Usually ships in 9 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
pH Meters and Water Quality Products
>
pH Meters, Ion Meters and ISFET Meters
>
pH/Ion Handheld Meters
>
Oakton pH 11 Handheld pH Meters
Share
|
Oakton pH 11 meter kit
EW-35614-80
$423.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
High quality and versatility at an economical price
Electrode bracket positions electrode for one-handed use
Built-in electrode stand allows for benchtop use
Data set storage, increased buffer recognition options, and auto-scanning buffer calibration
Basic pH 11 Meter features everything you need for general-purpose pH measurements with not-so-basic benefits. The custom, dual-LCD display provides measurement and troubleshooting data with its
decipherable icons. Recall up to 50 data sets and calibrate up to five pH points from USA and NIST buffers.
Large, dual-LCD lets you view pH (or mV) and temperature values simultaneously. Automatic temperature compensation (ATC) ensures accuracy even in fluctuating temperatures. Functions include temperature offset calibration, hold function, ready indicator, and auto-off. Electrode slope display capability allows for quick troubleshooting.
What's Included:
electrode bracket and batteries.
Meter with probe adds:
"All-in-One" pH electrode 35811-71 with built-in ATC
Meter kit further adds:
pH buffer (pH 4.01, 7.00, 10.00, and rinse water), solution bottles, and
hard carrying case.
Specifications
Product Type
pH Meter
Meter style
Portable
Dimensions (" W)
3-1/2
Dimensions (" H)
7-3/8
Dimensions (" D)
1-3/4
Range (pH)
2.00 to 16.00
Range (mV)
±1999
Range (Temperature °C)
10 to 110
Range (Temperature °F)
14 to 230
Resolution (pH)
0.1/0.01
Resolution (Temperature)
0.1
Resolution (mV)
0.1/1
Temperature compensation
Automatic or manual
Display
LCD
Calibration
Up to 5 points
Buffer recognition
USA, NIST
Power
Four AA batteries (included), optional universal AC adapter
Datalogging (points)
50
CE Compliance
Yes
Model
pH 11
Accessories
Qty
/
Price
Availability
Oakton buffer pack, 500-ml each of 4.01, 7.00, and 10.00 buffer
EW-05942-10
at
$29.00 USD/PACKAGE of 3
In stock.
Add to Cart
Electrode pH/Temp double-junction for high pH, low sodium error with 3-ft cable and ATC plug for Oakton pH5/6, 11/110, 510, 700, 1100/2100, 2700 meters
EW-05991-36
at
$202.00 USD/EACH
In stock.
Add to Cart
Batteries, 1.5 V, AAA, 12/pack
EW-09376-00
at
$21.25 USD/PACKAGE of 12
In stock.
Add to Cart
Oakton ATC probe for pH 5, pH 6, Ion 6, pH 11, pH 110, pH 510, Ion 510, pH 1100, and pH 2100 meters
EW-35613-05
at
$44.00 USD/EACH
In stock.
Add to Cart
Oakton adapter, 100/240 VAC 50/60 Hz
EW-35615-07
at
$55.00 USD/EACH
In stock.
Add to Cart
"All-in-one" combination, pH/Temp electrode, 3-ft cable, epoxy body, single-junction, gel filled, sub-miniature audio ATC, BNC, 12 mm dia.
EW-35811-71
at
$84.00 USD/EACH
In stock.
Add to Cart
"All-in-one" combination, pH/Temp electrode, 3-ft cable, epoxy body, double-junction, gel filled, sub-miniature audio ATC, BNC, 12 mm dia.
EW-35811-72
at
$124.00 USD/EACH
In stock.
Add to Cart
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; pH Meter
EW-17106-20
at $115.00 USD/EACH
Usually ships in 9 days.
Add to Cart
Technical Resources
Instruction Manual (1.13 MB)
Oakton Catalogo en Espanol.pdf (8.3 MB)
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
pH FAQs
Testing the pH Value of Pure Water
Testing the pH value of Meat
Show All...
Testing the pH in Yogurt Production
Testing the pH value of Wine
Testing the pH of Wastewater Samples
Testing the pH of Soil Samples
pH Electrode Theory
Oakton® pH meter becomes a TV star!
Measuring the pH of Pure Water
pH Systems
pH Temperature Compensation
Temperature Compensation for pH Instruments
pH Measurement FAQs
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Related media
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title