Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Oakton pH 11 meter with "All-in-One" electrode (35811-71)

EW-35614-20
$362.00 USD/EACH

  • High quality and versatility at an economical price
  • Electrode bracket positions electrode for one-handed use
  • Built-in electrode stand allows for benchtop use
  • Data set storage, increased buffer recognition options, and auto-scanning buffer calibration
Basic pH 11 Meter features everything you need for general-purpose pH measurements with not-so-basic benefits. The custom, dual-LCD display provides measurement and troubleshooting data with its
decipherable icons. Recall up to 50 data sets and calibrate up to five pH points from USA and NIST buffers.

Large, dual-LCD lets you view pH (or mV) and temperature values simultaneously. Automatic temperature compensation (ATC) ensures accuracy even in fluctuating temperatures. Functions include temperature offset calibration, hold function, ready indicator, and auto-off. Electrode slope display capability allows for quick troubleshooting.


What's Included:
electrode bracket and batteries.

Meter with probe adds: "All-in-One" pH electrode 35811-71 with built-in ATC

Meter kit further adds: pH buffer (pH 4.01, 7.00, 10.00, and rinse water), solution bottles, and
hard carrying case.
Product Type pH Meter
Meter style Portable
Dimensions (" W) 3-1/2
Dimensions (" H) 7-3/8
Dimensions (" D) 1-3/4
Range (pH) 2.00 to 16.00
Range (mV) ±1999
Range (Temperature °C) 10 to 110
Range (Temperature °F) 14 to 230
Resolution (pH) 0.1/0.01
Resolution (Temperature) 0.1
Resolution (mV) 0.1/1
Accuracy (pH) ±0.01 pH
Accuracy (Temperature) ±0.5°C
Accuracy (mV) ±0.2 mV or 0.05%, whichever is greater
Temperature compensation Automatic or manual
Display LCD
Calibration Up to 5 points
Buffer recognition USA, NIST
Power Four AA batteries (included), optional universal AC adapter
Datalogging (points) 50
CE Compliance Yes
Brand Oakton
Model pH 11

Accessories
Qty/Price
Availability

at $29.00 USD/PACKAGE of 3

In stock.

at $202.00 USD/EACH

In stock.

at $21.25 USD/PACKAGE of 12

In stock.

at $44.00 USD/EACH

In stock.

at $55.00 USD/EACH

In stock.

at $66.00 USD/EACH

In stock.

at $84.00 USD/EACH

In stock.

at $124.00 USD/EACH

In stock.
Calibrations
Qty/Price
Availability

at $115.00 USD/EACH
Usually ships in 9 days.




Technical Resources
 
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title