Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Oakton pH 300 meter with single-junction pH/ATC probe

EW-35618-03
$572.00 USD/EACH

  • Waterproof and dustproof with IP67-rated housing that floats!
  • Built-in memory function makes it easy to recall data
  • Five-point pH calibration with auto-buffer recognition
Basic pH 300 Meter functions include mV and temperature offset calibration, HOLD function, Ready indicator, and auto-off. Memory stores 16 readings, recalls electrode slope and offset, and previous calibration data.

What's Included:
two electrode brackets and batteries.

Meter with probe adds: ''All-in-One'' pH electrode with built-in ATC.

Meter kit further adds: pH buffer (pH 4.01, 7.00, 10.01, and rinse water), sample bottles, and hard carrying case.
Product Type pH Meter
Meter style Portable
Dimensions (" W) 3-1/2
Dimensions (" H) 7-3/8
Dimensions (" D) 1-3/4
Range (pH) 2.00 to 16.00
Range (mV) ±1999
Range (Temperature °C) 10 to 110
Range (Temperature °F) 14 to 230
Resolution (pH) 0.1/0.01
Resolution (Temperature) 0.1
Resolution (mV) 0.1/1
Accuracy (pH) ±0.01 pH
Accuracy (Temperature) ±0.5°C
Accuracy (mV) ±0.2 mV or 0.05%, whichever is greater
Temperature compensation Automatic or manual
Display LCD
Calibration Up to 5 points
Waterproof Yes
Buffer recognition USA, NIST, DIN
Power Four AAA batteries (included)
Datalogging (points) 16
CE Compliance Yes
Brand Oakton
Model pH 300

Accessories
Qty/Price
Availability

at $29.00 USD/PACKAGE of 3

In stock.

at $59.00 USD/EACH

In stock.

at $66.00 USD/EACH

In stock.

at $109.00 USD/EACH

In stock.

at $127.00 USD/EACH

In stock.

at $117.00 USD/EACH

In stock.
Calibrations
Qty/Price
Availability

at $115.00 USD/EACH
Usually ships in 9 days.
Reconditioned & Like New
Qty/Price
Availability
LIKE NEW 35618-03
EW-00173ZR

at $464.10 USD/EACH

In stock.




Technical Resources
 
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title