United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 5
of
8 >
EW-35419-10
pH 700 meter
Click to enlarge
More Product Details
Specifications
Accessories
Calibrations
Technical Resources
Technical information
Instruction manual
Virtual Catalog (PDFs)
More info
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-35419-10
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17106-20
NIST-Traceable Calibration; pH Meter
$115.00 USD/EACH/EACH
Usually ships in 9 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
pH Meters and Water Quality Products
>
pH Meters, Ion Meters and ISFET Meters
>
pH/Ion Benchtop Meters
>
Oakton Benchtop pH/Ion Meters
>
Oakton Benchtop pH 700 Meter
Share
|
Oakton pH 700 Benchtop Meter, double-junction refillable glass pH electrode, ATC probe and stand
EW-35419-10
$631.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Designed for today's crowded laboratory benchtops!
Compact footprint is over 40% smaller than other benchtop meters
Large display permits easier readout viewing
Datalogging allows you to store and recall up to 100 data sets
The easy-to-use, microprocessor-based pH 700 benchtop meter measures pH, mV, relative mV, and temperature. The large, dual-display LCD provides excellent visibility—even from a distance. Automatic temperature compensation (ATC) maintains reading accuracy even with fluctuating temperatures. Five-point pH calibration, with automatic buffer recognition for both USA (pH 1.68, 4.01, 7.00, 10.01, 12.45) and NIST (pH 1.68, 4.01, 6.86, 9.18, 12.45) pH buffers, makes for easy calibration no matter which requirement you need to meet.
Meter features HOLD, ATC, selectable °f or °C measurement, recall of electrode slopand offset, and electrode diagnostics. The convenient pull-out quick reference card keeps procedures handy at all times. Meter accepts any pH or ORP electrodes with a BNC connector.
What's Included:
universal 110/240 VAC power supply and adapters.
Specifications
Product Type
pH Meter
Meter style
Benchtop
Dimensions (" W)
6-7/8
Dimensions (" H)
2-3/4
Dimensions (" L)
6-1/8
Range (pH)
2.00 to 16.00
Range (mV)
± 2000
Range (Temperature °C)
10 to 110
Range (Temperature °F)
14 to 230
Resolution (pH)
0.1/0.01
Resolution (Temperature)
0.1
Resolution (mV)
0.1/1
Accuracy (pH)
±0.01 pH
Accuracy (Temperature)
±0.5°C
Accuracy (mV)
±0.2 mV or 0.05%, whichever is greater
Temperature compensation
Automatic or manual
Display
LCD
Calibration
Up to 6 points
Connections
BNC, Temp 2.5mm
Buffer recognition
USA, NIST
Power
100 to 240 VAC, 50/60 Hz
Datalogging (points)
100
CE Compliance
Yes
Brand
Oakton
Model
pH 700
Accessories
Qty
/
Price
Availability
Oakton buffer pack, 500-ml each of 4.01, 7.00, and 10.00 buffer
EW-05942-10
at
$29.00 USD/PACKAGE of 3
In stock.
Add to Cart
Electrode pH/Temp double-junction for high pH, low sodium error with 3-ft cable and ATC plug for Oakton pH5/6, 11/110, 510, 700, 1100/2100, 2700 meters
EW-05991-36
at
$202.00 USD/EACH
In stock.
Add to Cart
Replacement Electrode Arm for Oakton Meters
EW-35420-54
at
$77.00 USD/EACH
In stock.
Add to Cart
Replacement temperature probe
EW-35613-13
at
$43.50 USD/EACH
In stock.
Add to Cart
Oakton pH electrode, glass body, refillable, double-junction, 3-ft (0.9-m) cable, BNC connector
EW-35805-04
at
$128.00 USD/EACH
In stock.
Add to Cart
"All-in-one" combination, pH/Temp electrode, 3-ft cable, epoxy body, single-junction, gel filled, sub-miniature audio ATC, BNC, 12 mm dia.
EW-35811-71
at
$84.00 USD/EACH
In stock.
Add to Cart
pH electrode, combination, single-junction, gel-filled, BNC connector
EW-59001-65
at
$70.00 USD/EACH
In stock.
Add to Cart
pH electrode, combination, double-junction, gel-filled, BNC connector
EW-59001-70
at
$84.00 USD/EACH
In stock.
Add to Cart
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; pH Meter
EW-17106-20
at $115.00 USD/EACH
Usually ships in 9 days.
Add to Cart
Technical Resources
Quick start guide (English)
Quick start guide (Portuguese)
Instruction Manual for 35419-10.pdf
Virtual Catalog (PDFs)
13-14 Catalog
More info
700 Series Quick Guide_German
700 Series Quick Guide_Spanish
Related technical articles (Roll over for summary)
pH FAQs
Testing the pH Value of Pure Water
Testing the pH value of Meat
Show All...
Testing the pH in Yogurt Production
Testing the pH value of Wine
Testing the pH of Wastewater Samples
Testing the pH of Soil Samples
pH Electrode Theory
Oakton® pH meter becomes a TV star!
Measuring the pH of Pure Water
pH Systems
pH Temperature Compensation
Temperature Compensation for pH Instruments
pH Measurement FAQs
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title