Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Oakton pH 700 Benchtop Meter, meter and probe stand

EW-35419-12
$518.00 USD/EACH

  • Designed for today's crowded laboratory benchtops!
  • Compact footprint is over 40% smaller than other benchtop meters
  • Large display permits easier readout viewing
  • Datalogging allows you to store and recall up to 100 data sets
The easy-to-use, microprocessor-based pH 700 benchtop meter measures pH, mV, relative mV, and temperature. The large, dual-display LCD provides excellent visibility—even from a distance. Automatic temperature compensation (ATC) maintains reading accuracy even with fluctuating temperatures. Five-point pH calibration, with automatic buffer recognition for both USA (pH 1.68, 4.01, 7.00, 10.01, 12.45) and NIST (pH 1.68, 4.01, 6.86, 9.18, 12.45) pH buffers, makes for easy calibration no matter which requirement you need to meet.

Meter features HOLD, ATC, selectable °f or °C measurement, recall of electrode slopand offset, and electrode diagnostics. The convenient pull-out quick reference card keeps procedures handy at all times. Meter accepts any pH or ORP electrodes with a BNC connector.Order pH electrode separately.
What's Included:
universal 110/240 VAC power supply and adapters.
Product Type pH Meter
Meter style Benchtop
Dimensions (" W) 6-7/8
Dimensions (" H) 2-3/4
Dimensions (" L) 6-1/8
Range (pH) 2.00 to 16.00
Range (mV) ± 2000
Range (Temperature °C) 10 to 110
Range (Temperature °F) 14 to 230
Resolution (pH) 0.1/0.01
Resolution (Temperature) 0.1
Resolution (mV) 0.1/1
Accuracy (pH) ±0.01 pH
Accuracy (Temperature) ±0.5°C
Accuracy (mV) ±0.2 mV or 0.05%, whichever is greater
Temperature compensation Automatic or manual
Display LCD
Calibration Up to 6 points
Connections BNC, Temp 2.5mm
Buffer recognition USA, NIST
Power 100 to 240 VAC, 50/60 Hz
Datalogging (points) 100
CE Compliance Yes
Brand Oakton
Model pH 700

Accessories
Qty/Price
Availability

at $29.00 USD/PACKAGE of 3

In stock.

at $202.00 USD/EACH

In stock.

at $77.00 USD/EACH

In stock.

at $43.50 USD/EACH

In stock.

at $128.00 USD/EACH

In stock.

at $84.00 USD/EACH

In stock.

at $70.00 USD/EACH

In stock.

at $84.00 USD/EACH

In stock.
Calibrations
Qty/Price
Availability

at $115.00 USD/EACH
Usually ships in 9 days.




Technical Resources
 
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title