United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
Representative photo only
Oakton pH Solution Pack one 500 mL bottle each of pH 4 01 7 00 10 00 storage and cleaning solution
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
MSDS
Brochure
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-05942-15
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
pH Meters and Water Quality Products
>
Buffers, Standards, and Solutions
>
pH/Ion Buffers, and Storage Solutions
>
Buffers and Solutions for pH/Ion Measurement
>
Oakton pH Solution Five Pack
Share
|
Oakton pH Solution Pack, one 500-mL bottle each of pH 4.01, 7.00, 10.00, storage, and cleaning solution
EW-05942-15
$53.50 USD/PACKAGE
In stock.
Qty
:
Add to Cart
Save to shopping list
Save over 25% compared to ordering separately!
Economical and convenient way to order everything you need to calibrate, clean, and store your pH electrodes. Pack includes one 500-mL bottle each of pH 4.01, 7.00, and 10.00 buffers; one 500-mL bottle of cleaning solution for grease, oil, and protein removal; and one 500-mL bottle of storage solution to properly store your electrode while not in use.
Specifications
Product Type
pH Buffers
Volume (mL)
500
Quantity
500mL
Brand
Oakton
Manufacturer number
05942-10,00653-06 & -04
Technical Resources
MSDS
Oakton Catalogo en Espanol.pdf (8.3 MB)
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Tips for pH Buffer Use
Testing the pH of Wastewater Samples
Testing the pH of Soil Samples
Show All...
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title