Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
 

Oakton pH Solution Pack, one 500-mL bottle each of pH 4.01, 7.00, 10.00, storage, and cleaning solution

SI-05942-15
$53.50 USD/PACKAGE

  • Save over 25% compared to ordering separately!
Economical and convenient way to order everything you need to calibrate, clean, and store your pH electrodes. Pack includes one 500-mL bottle each of pH 4.01, 7.00, and 10.00 buffers; one 500-mL bottle of cleaning solution for grease, oil, and protein removal; and one 500-mL bottle of storage solution to properly store your electrode while not in use.
Product Type pH Buffers
Volume (mL) 500
Quantity 500mL
Brand Oakton
Manufacturer number 05942-10,00653-06 & -04





Technical Resources
 
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title