Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Oakton pH buffer pouches; pH 7.00, 20/pk

EW-35653-02
$31.90 USD/PACKAGE of 20

  • Economical pouches are accurate and convenient
Buffers are NIST-traceable and offer ±0.01 accuracy at 77°F (25°C). Pack of 20 pouches.
Product Type pH Buffers
Description pH 7.00
Quantity 20 pouches
Qty/pk 20
Brand Oakton
Manufacturer number 35653-02





Technical Resources
Certificate of Analysis
Enter lot number

or click to view a Sample C of A
 
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title