United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 5
of
15 >
Representative photo only
Oakton pH electrode epoxy body sealed double junction general purpose 3 ft 0 9 m cable BNC connector
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Brochure
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-35805-01
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
pH Meters and Water Quality Products
>
Electrodes, Laboratory
>
pH/Ion Laboratory Electrodes
>
Oakton Combination pH Electrodes
Share
|
Oakton pH electrode, epoxy-body, sealed, double-junction, general purpose, 3-ft (0.9-m) cable, BNC connector
EW-35805-01
$73.44 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Outstanding value at an economical cost!
Compatible with all Oakton brand pH meters
Oakton combination electrodes are available in a variety of styles to meet the needs of your application. Epoxy electrodes are impact resistant and provide exellent durability for field applications. Glass electrodes withstand aggressive chemicals. Choose junction and tip style based on the needs of your application. All electrodes feature Ag/AgCl reference cells.
What's Included:
direct-connect models include a BNC connector. Other models add a 3.3-ft (1-m) cable.
Specifications
Product Type
pH Electrode
Body type
Epoxy
Reference junction
Double-junction
Special features
General purpose
pH range
0 to 12
Temperature range
32 to 176°F (0 to 80°C)
Reference cell type
Ag/AgCl
Cable length
3-ft (0.9-m)
Connector
BNC
Built-in ATC
No
Brand
Oakton
Technical Resources
Oakton Catalogo en Espanol.pdf (8.3 MB)
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
pH FAQs
Spectroscopy Applications Selection Guide
Testing the pH Value of Pure Water
Show All...
Testing the pH value of Meat
Testing the pH in Yogurt Production
Testing the pH value of Wine
Part 2: Troubleshooting pH Electrodes
Series Part 1: How to Store, Clean, and Recondition pH Electrodes
pH Electrode Selection Guide
pH Electrode Theory
Measuring the pH of Pure Water
pH Systems
pH Electrode Care
pH Measurement FAQs
Laboratory pH Electrodes
Testing the pH of Wastewater Samples
Testing the pH of Soil Samples
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title