United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
2 >
EW-01000-00
Ohaus Cent-O-Gram Overhead Mechanical Balance, 311 g x 0.01 g
Click to enlarge
More Product Details
Specifications
Accessories
Related products
Calibrations
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-01000-00
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17101-52
NIST Traceable Report With Data, Balances/Scales - all types
$140.00 USD/EACH/EACH
Usually ships in 7 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Balances & Scales
>
Mechanical Balances
>
Ohaus Overhead Mechanical Balances
Share
|
Ohaus Cent-O-Gram Overhead Mechanical Balance, 311 g x 0.01 g
EW-01000-00
$238.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
:
Add to Cart
Save to shopping list
Get stable readings three times faster than other three-beam balances
Designed with a hanging pan system
Offer 10 times the readability of standard triple-beam balances
Feature a self-aligning beam, floating agate bearings, and counter-balancing knob for fast zeroing
Dial-O-Gram® balance comes equipped with a convenient, direct-reading dial calibration
Magnetic damping speeds up weighing times
Specifications
Product Type
Mechanical Balances
Balance type
Cent-O-Gram® four-beam
Beam calibrations
front beam: 1g x 0.01g, 2nd beam: 10g x 1g, 3rd beam: 100g x 10g, rear beam: 200g x 100g)
Calibration
External
Capacity (g)
311
Capacity (kg)
0.31
Capacity (oz)
11
Capacity (lbs)
0.68
Housing dimensions
35.5 x 24 x 17.8 cm
Dimensions (" W)
14
Dimensions (" H)
9
Dimensions (" D)
7
Pan size (dia)
3.5" x 0.5"D (9 x 0.7 cm)
Readability (g)
0.01
Linearity (g)
mechanical
Repeatability (g)
mechanical
Weighing units
g
Brand
Ohaus
Model
311-00
Accessories
Qty
/
Price
Availability
Vinyl Dust Cover for Ohaus Cent-O-Gram and Dial-O-Gram balances (01000-00 and 01330-00)
EW-01003-00
at
$22.00 USD/EACH
Usually ships in 6 days.
Add to Cart
Related products
Qty
/
Price
Availability
Cole-Parmer SpatulaBalance
, 300 g x 0.1 g
EW-01019-11
at $43.00 USD/EACH
In stock.
Add to Cart
Calibrations
Qty
/
Price
Availability
NIST Traceable Report With Data, Balances/Scales - all types
EW-17101-52
at $140.00 USD/EACH
Usually ships in 7 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Gravitational Effects on Balance
Weighing Unit Conversion Table
Common Balance Terms
Show All...
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title