United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 4
of
4
Representative photo only
Ohaus Compact Thermal Printer
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-11901-47
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Balances & Scales
>
Balances / Scales Accessories
>
Ohaus Balance Accessories
>
Ohaus Printers
>
Ohaus Compact Printers
Share
|
Ohaus Compact Thermal Printer
EW-11901-47
$467.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Connects to your RS-232 interface for accurate record keeping
Lightweight and compact
Never needs a printer ribbon
Easy paper loading
Small footprint: 100 (W) x 191 (D) x 90 (H) mm
For Voyager Pro and Explorer Pro balances, order RS232 cable 11901-54
For Discovery, Adventurer Pro, Pioneer, Moisture Balances MB35/MB45, Ranger, Ranger Count, and T Series Dry Indicators, order RS232 cable 11901-55
For Trooper series, order RS232 cable 11901-56
For Ohaus CW, Champ CKW, and T Series Washdown Indicators, order RS232 cable 11901-57
For Ohaus Scout Pro balances, order RS232 adapter 11901-58
What's Included:
AC power adapter, power cord, and one roll of paper. Order additional roll of paper 11901-53 separately.
Specifications
Product Type
Ohaus Compact Printers
Brand
Ohaus
Manufacturer number
80251992
Model
80251992
Accessories
Qty
/
Price
Availability
Ohaus Compact Thermal Printer Paper
EW-11901-53
at
$11.00 USD/EACH
Usually ships in 7 days.
Add to Cart
OHAUS RS-232 CABLE AND ADAPTER FOR COMPACT THERMAL PRINTER 11901-47
EW-11901-54
at
$115.00 USD/EACH
Usually ships in 7 days.
Add to Cart
OHAUS RS-232 CABLE AND ADAPTER FOR COMPACT THERMAL PRINTER 11901-47
EW-11901-55
at
$115.00 USD/EACH
Usually ships in 7 days.
Add to Cart
OHAUS RS-232 CABLE AND ADAPTER FOR COMPACT THERMAL PRINTER 11901-47
EW-11901-56
at
$115.00 USD/EACH
Usually ships in 7 days.
Add to Cart
OHAUS RS-232 CABLE AND ADAPTER FOR COMPACT THERMAL PRINTER 11901-47
EW-11901-57
at
$128.00 USD/EACH
In stock.
Add to Cart
Ohaus RS-232 Cable Adapter For Compact Thermal Printer 11901-47
EW-11901-58
at
$19.00 USD/EACH
Usually ships in 7 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Weighing Unit Conversion Table
Common Balance Terms
Equipping a Laboratory: A Study in Customer Service Excellence
Show All...
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title