United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 3
of
3
EW-11800-02
Ohaus Explorer® Analytical Balance, 320g X 0.0001g. Representative Photo Only
Click to enlarge
More Product Details
Specifications
Accessories
Calibrations
Technical Resources
Virtual Catalog (PDFs)
More info
Related technical articles
Service & Support
Related media
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-11800-02
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17101-52
NIST Traceable Report With Data, Balances/Scales - all types
$140.00 USD/EACH/EACH
Usually ships in 7 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Balances & Scales
>
Analytical Balances
>
Ohaus Analytical Balances
>
Ohaus Explorer Analytical Balances
Share
|
Ohaus Explorer Analytical Balance, 320g X 0.0001g
EW-11800-02
$4,995.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Ingenious modular design with improved performance
Modular design allows for the display to be separated from the weighing base for maximum flexibility in any work area
AutoCal™ automatic internal calibration ensures results are accurate
Intuitive operation with large color touch-screen display and icon-driven software
Easy-to-use SmarText™ graphical software
Touchless sensors for hands-free operation
The OHAUS Explorer’s advanced applications simplify even the most complex laboratory measurements. Whether it’s determining the difference between initial and residual weights or calculating the density of solids and liquids, Explorer eliminates the need for time consuming manual calculations and data logging. The high resolution display and innovative user interface make balance setup and application use effortless. Features include
Intelligent Performance – All Explorers have newly optimized linearity and repeatability specifications and enhanced vibration filtering for better balance stability. All models also come standard with AutoCal™ automatic internal calibration. These combined enhancements improve accuracy, efficiency, and throughput.
• Intuitive Operation – A large color touch-screen display, icon-driven application software that features 14 unique application modes, and an adjustable angle display make Explorer the easiest to use, most advanced balance in its class today.
• Ingenious Design – The base and terminal separate for maximum flexibility in any work area. The design also isolates the weighing chamber to enhance stability and speed results. Draftshield models feature an oversized chamber with a new quick remove design, enhanced with a unique anti static system.
• Practical Features – Explorer has four user-programmable touch free sensors to minimize cross contamination and button wear. All Explorers meet or exceed the Class I and II accuracy requirements in accordance with the National Institute of Standards and Technology (NIST) Handbook 44 (approval pending).
• Intuitive SOFTWARE - SmarText™ 2.0 is OHAUS’ easy-to-use graphical software featuring 14 applications, QWERTY / numeric keypads, and below-minimum weight indication.
• Ingenious DRAFTSHIELD - Explorer’s draftshield provides ample access and visibility to the weighing chamber and features antistatic coated glass.
• Practical TOUCHLESS SENSORS - Explorer features four touchless sensors for hands-free operation of print, calibration, tare, and other selectable functions.
• Intelligent CALIBRATION - AutoCal™ ensures performance and assists with routine maintenance by automatically calibrating the balance daily.
• Intuitive USER SETUP - Explorer is the industry’s most easy-to-use balance, featuring leveling assistance and instructional messaging for quick out-of-the-box setup and use.
• Ingenious MODULAR DESIGN - Explorer’s modular design features a color touch display that can be separated from the weighing base.
• Practical APPROVALS - Explorer is designed and engineered to meet stringent NTEP requirements for use in legal applications such as pharmacies, jewelry stores and retail outlets.
What's Included:
Below-balance weighing hook, AC Adapter, security bracket, and draft shield
Specifications
Product Type
Analytical Balances
Calibration
Internal
Capacity (g)
320
Capacity (oz)
11
Display
Full-color VGA graphic display, 4-wire resistive touch screen
Pan size (dia)
3-1/2" (8.9 cm)
Readability (g)
0.0001
Linearity (g)
±0.0002
Repeatability (g)
±0.0001
Weighing units
g, mg, kg, ct, oz, dwt, gn, lb tael
Stabilization time
3 sec
Stability filters
selectable environmental filters
Power (VAC)
100/240
Languages
English German French Spanish Italian Russian Polish Chinese Portuguese Korean Japanese
Output
RS-232 / USB / auxiliary
Power
50/60 Hz
Brand
Ohaus
Model
EX324
Accessories
Qty
/
Price
Availability
Ohaus Explorer Tower Mount for Display
EW-11800-13
at
$99.00 USD/EACH
Usually ships in 4 days.
Add to Cart
Ohaus Explorer Display Extension Cable 10 foot
EW-11800-14
at
$52.00 USD/EACH
Usually ships in 4 days.
Add to Cart
Ohaus Explorer Density Determination Kit
EW-11800-15
at
$469.00 USD/EACH
Usually ships in 4 days.
Add to Cart
Ohaus Explorer Draftshield Kit for 0.01g & 0.1g models
EW-11800-16
at
$567.00 USD/EACH
Usually ships in 4 days.
Add to Cart
Ohaus Explorer Interface Cable, USB (Type A to B)
EW-11800-18
at
$115.00 USD/EACH
In stock.
Add to Cart
Ohaus Explorer Second RS232 Kit
EW-11800-19
at
$115.00 USD/EACH
Usually ships in 4 days.
Add to Cart
Ohaus Explorer Ethernet Kit
EW-11800-20
at
$115.00 USD/EACH
Usually ships in 4 days.
Add to Cart
Ohaus Explorer Foot Switch Kit
EW-11800-21
at
$110.00 USD/EACH
Usually ships in 4 days.
Add to Cart
Ohaus Explorer Compact Thermal Printer
EW-11800-22
at
$467.00 USD/EACH
Usually ships in 4 days.
Add to Cart
Ohaus Explorer Compact Dot Matrix Impact Printer
EW-11800-23
at
$549.00 USD/EACH
Usually ships in 4 days.
Add to Cart
Ohaus Explorer RS232 Cable & Adapter to Compact Thermal Printer
EW-11800-24
at
$115.00 USD/EACH
Usually ships in 4 days.
Add to Cart
Ohaus Explorer RS232 Cable & Adapter to Compact Dot Matrix Impact Printer
EW-11800-25
at
$115.00 USD/EACH
Usually ships in 4 days.
Add to Cart
Ohaus Explorer RS232 Cable, PC 25 Pin to EX
EW-11800-26
at
$115.00 USD/EACH
Usually ships in 4 days.
Add to Cart
Ohaus Explorer RS232 Cable, PC 9 Pin to EX
EW-11800-27
at
$115.00 USD/EACH
Usually ships in 4 days.
Add to Cart
Calibrations
Qty
/
Price
Availability
NIST Traceable Report With Data, Balances/Scales - all types
EW-17101-52
at $140.00 USD/EACH
Usually ships in 7 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
More info
Tektronix MSO/DPO Series sell sheet (651K)
Related technical articles (Roll over for summary)
Gravitational Effects on Balance
Weighing Unit Conversion Table
Common Balance Terms
Show All...
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Related media
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title