United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 7
of
12 >
EW-01002-84
Ohaus MB Moisture Determination Balance, Mb45 45 G X 0.001 G 120VAC
Click to enlarge
More Product Details
Specifications
Accessories
Related products
Calibrations
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-01002-84
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17101-52
NIST Traceable Report With Data, Balances/Scales - all types
$140.00 USD/EACH/EACH
Usually ships in 7 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Balances & Scales
>
Moisture Balances
>
Ohaus MB Moisture Balances
Share
|
Ohaus MB Moisture Determination Balance, Mb45 45 gX 0.001 G 120VAC
EW-01002-84
$4,195.00 USD/EACH
Usually ships in 12 days.
Qty
:
Add to Cart
Save to shopping list
Versatile and rugged design for continuous operation in food and beverage, quality control, environmental, and many other applications
Choice of infrared or halogen heat source and programmable operation satisfy a wider variety of applications
Durable, compact construction ensures dependable, long-term operation within minimum benchtop space
Straightforward operation speeds up routine testing applications and accommodates operators of all skill levels
Sample pan holder included.
Ohaus MB Series moisture analyzers are the quick, effective, and affordable solution to measure moisture content. These moisture analyzers cover a wider variety of weighing applications in the pharmaceutical, chemical and research industries with excellent repeatability properties for consistent, accurate testing.
MB 45 Advanced Moisture Analyzers
handle the most demanding applications. An integrated database stores up to 50 test procedures to maximize your productivity simply recall your frequently used test from the database and begin analyzing your sample. Advanced moisture analyzers offer a maximum weighing capacity of 45 g, and four automatic drying programs for easy one step drying simply set the desired temperature and time and begin. A built-in statistical function provides simple tracking and recording of standard deviation over time, performs all calculations and provides the end result. The bright LCD shows % moisture, % solids, weight, time, temperature, and drying curve in English, Spanish, French, Italian, and German languages.
What's Included:
50 sample pans, fiber pads, in-use cover, and 6-ft (1.8 m) power cord
Specifications
Product Type
Moisture Balances
Calibration
External
Capacity (g)
45
Capacity (oz)
1.6
Heater type
Halogen
Housing dimensions
7-1/2"W x 6"H x 14-1/4"D (19.1 x 15.2 x 36.2 cm)
Pan size (dia)
3-1/2" (8.9 cm)
Power (VAC)
120
Readability
0.001 g
Repeatability
±0.05% (3 g sample)
Temperature range (º C) (low)
50
Temperature range (º C) (high)
200
Timer (min)
1 to 120
Weighing units
% moisture, % solids, % regain, statistics, time, temperature, weight (g), test ID, drying curve
Output
RS-232
Brand
Ohaus
Accessories
Qty
/
Price
Availability
Aluminum Pan Liners, 3.3gr, 127mm i.d.(5") x 3/16" depth, 1000/cs
EW-01018-22
at
$171.50 USD/CASE of 1000
In stock.
Add to Cart
Ohaus Compact Thermal Printer
EW-11901-47
at
$467.00 USD/EACH
In stock.
Add to Cart
Ohaus Compact Thermal Printer Paper
EW-11901-53
at
$11.00 USD/EACH
Usually ships in 7 days.
Add to Cart
OHAUS RS-232 CABLE AND ADAPTER FOR COMPACT THERMAL PRINTER 11901-47
EW-11901-55
at
$115.00 USD/EACH
Usually ships in 7 days.
Add to Cart
Ohaus Compact Dot Matrix Printer
EW-11901-59
at
$549.00 USD/EACH
Usually ships in 7 days.
Add to Cart
Ohaus Compact Dot Matrix Printer Paper For 11901-59
EW-11901-60
at
$10.00 USD/EACH
Usually ships in 7 days.
Add to Cart
Ohaus RS-232 Cable And Adapter For Compact Dot Matrix Printer 11901-59 f/moisture bal
EW-11901-63
at
$115.00 USD/EACH
Usually ships in 7 days.
Add to Cart
Related products
Qty
/
Price
Availability
Cole-Parmer SpatulaBalance
, 300 g x 0.1 g
EW-01019-11
at $43.00 USD/EACH
In stock.
Add to Cart
Calibrations
Qty
/
Price
Availability
NIST Traceable Report With Data, Balances/Scales - all types
EW-17101-52
at $140.00 USD/EACH
Usually ships in 7 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Taking the “Dry” Out of Moisture Determination
Gravitational Effects on Balance
Weighing Unit Conversion Table
Show All...
Common Balance Terms
Equipping a Laboratory: A Study in Customer Service Excellence
Temperature FAQs
Temperature Instrument Ranges
Getting a Handle on Temperature
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title